Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00488345_m1
          See other SNCG GE Assays ›
          Gene Symbol:
          Sncg
          Gene Name:
          synuclein, gamma
          Gene Aliases:
          C79089, persyn
          Chromosome Location:
          Chr.14: 34370274 - 34374669 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00488345_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Sncg
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_011430.3 NP_035560.1 3 - 4 382 93
          GenBank mRNA AF017255.1 - 3 - 4 361 93
          AJ786826.1 - 3 - 4 295 93
          AK141509.1 - 3 - 4 366 93
          BC028508.1 - 3 - 4 373 1448798 93

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1400001725.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Mus musculus Mouse Tumor Metastasis Panel 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Sncg

          Panther Classification:

          Molecular Function -

          membrane trafficking regulatory protein

          Gene Ontology Categories:

          Function(s) Process(es)

          chemical synaptic transmission
          adult locomotory behavior
          protein secretion
          regulation of dopamine secretion
          regulation of neurotransmitter secretion
          synapse organization
          protein binding
          alpha-tubulin binding
          beta-tubulin binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2187076 NM_011430.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2187076 AK141509.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2187076 BC028508.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 139824 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 712648 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 689345 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 438581 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 18002 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P120879 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2187076 NM_011430.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2187076 AK141509.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2187076 BC028508.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P118300 NM_011430.3 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P118300 AA690669.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P118300 AF017255.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P118300 AJ786826.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P118300 AK141509.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29529625 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29529624 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29529604 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29529616 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29529609 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29529605 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29529619 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29529608 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29529638 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29529607 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29529633 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29529634 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29529602 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29529626 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29529623 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29529640 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29529612 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29529611 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29529620 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29529628 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29529644 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29529630 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29529635 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29529615 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29529642 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29529621 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29529613 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P120879 - -
          Whole Mouse Genome G4122A A_51_P118300 NM_011430.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P118300 AA690669.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P118300 AF017255.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P118300 AJ786826.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P118300 AK141509.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P139099 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P120879 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11408063 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11408069 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11408077 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11408062 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11408076 - -
          Agilent Mouse cDNA Microarray (G4104A) 1195168 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29529588 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29529550 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29529564 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29529611 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29529591 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29529621 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29529581 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29529635 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29529603 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29529607 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29529604 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29529633 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29529577 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29529625 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29529597 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29529630 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29529583 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29529619 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29529563 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29529575 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29529612 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29529576 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29529594 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29529615 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29529626 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29529608 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29529640 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29529571 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29529644 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29529554 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29529623 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29529613 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29529592 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29529624 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29529572 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29529628 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29529638 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29529596 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29529561 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29529567 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29529553 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29529620 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29529642 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29529585 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29529598 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29529549 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29529605 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29529555 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29529589 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29529562 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29529587 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29529574 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29529569 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29529633 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29529611 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29529598 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29529620 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29529608 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29529596 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29529603 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29529613 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29529635 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11763721 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29529640 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29529621 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29529617 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11603878 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11763725 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29529594 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29529623 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29529638 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29529619 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29529601 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11235253 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11763744 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29529630 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29529615 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11314225 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29529624 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29529607 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10418924 - -
          Affymetrix Mouse Gene 1.0 ST Array 10418925 - -
          Affymetrix Mouse Gene 1.0 ST Array 10418922 - -
          Affymetrix Mouse Gene 1.0 ST Array 10418923 - -
          Affymetrix Mouse Gene 1.0 ST Array 10418926 - -
          Affymetrix HT MG-430 PM Array Plate 1417788_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10418921 - -
          Mouse Genome 430 2.0 Array 1417788_at - -
          Mouse Genome 430 2.0 Array 1417788_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 20618_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 20618_at - -
          Affymetrix Mouse Clariom S Array PSR1400013423.mm.1 NM_011430.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400013423.mm.1 NM_011430.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1400013423.mm.1 TC1400001725.mm.1 MTA TC
          Mouse Expression Set 430 1417788_at - -
          Mouse Expression Set 430 1417788_PM_at - -
          Murine Genome U74v2 Set 161859_PM_f_at - -
          Murine Genome U74v2 Set 161859_f_at - -
          Murine Genome U74v2 Set 104280_PM_at - -
          Murine Genome U74v2 Set 104280_at - -
          Mouse Genome 430A 2.0 Array 1417788_PM_at - -
          Mouse Genome 430A 2.0 Array 1417788_at - -
          Affymetrix HT Mouse Genome 430A Array 1417788_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1417788_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 796589 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 3120725 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003120725 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2598478 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2598478 - -
          mouse-6 v1.1 (Illumina) scl45723.1.148_13-S - -
          Illumina Sentrix mouse-6.v1 bead array 3120725 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl45723.1.148_13-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3120725 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2598478 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2939277 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2939277 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2598478 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl45723.1.148_13-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline