Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00488528_m1
          See other ALYREF GE Assays ›
          Gene Symbol:
          Alyref
          Gene Name:
          Aly/REF export factor
          Gene Aliases:
          ALY, REF1, Refbp1, Thoc4
          Chromosome Location:
          Chr.11: 120594516 - 120598436 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00488528_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Alyref
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_011568.1 NP_035698.1 1 - 2 264 115
          GenBank mRNA BC120588.1 - 1 - 2 356 40129511 115
          BC137658.1 - 1 - 2 356 8860677 115
          U89876.1 - 1 - 2 264 115

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1100004219.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Alyref

          Panther Classification:

          Molecular Function -

          RNA metabolism protein

          Gene Ontology Categories:

          Function(s) Process(es)

          mRNA processing
          transport
          RNA splicing
          mRNA transport
          nucleotide binding
          nucleic acid binding
          single-stranded DNA binding
          RNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28185534 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28185530 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28185532 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28185529 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28185531 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P154684 U89876.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P154684 NM_011568.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P154684 AK035721.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P154684 U89876.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P154684 NM_011568.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P154684 AK035721.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 79110 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 897794 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 314498 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 159643 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P146668 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P154684 U89876.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P154684 NM_011568.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P154684 AK035721.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P154684 U89876.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P154684 NM_011568.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P154684 AK035721.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P154684 U89876.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P154684 NM_011568.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P154684 AK035721.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P154684 U89876.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P154684 NM_011568.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P154684 AK035721.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28185498 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28185522 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28185493 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28185525 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28185503 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28185516 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28185495 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28185501 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28185511 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28185526 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28185523 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28185519 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28185529 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28185515 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28185530 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28185512 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28185521 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28185532 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28185506 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28185528 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28185514 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07516796 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P146668 - -
          Whole Mouse Genome G4122A A_51_P154684 U89876.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P154684 NM_011568.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P154684 AK035721.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P154684 U89876.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P154684 NM_011568.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P154684 AK035721.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P111616 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P138049 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P146668 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09396573 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28185530 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28185529 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28185534 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28185531 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28185532 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28185530 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28185498 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28185523 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28185514 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28185490 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28185487 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28185528 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28185459 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28185516 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28185463 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28185515 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28185472 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28185460 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28185489 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28185506 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28185495 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28185491 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28185512 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28185500 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28185529 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28185457 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28185462 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28185493 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28185521 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28185467 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28185473 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28185526 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28185502 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28185485 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28185512 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28185506 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28185519 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09340811 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09534181 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28185523 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28185500 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28185516 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28185493 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28185529 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09534168 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28185521 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28185526 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28185491 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28185485 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28185524 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09396579 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28185487 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28185509 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28185495 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28185498 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09534175 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09735366 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28185530 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28185490 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28185502 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28185528 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10393849 - -
          Affymetrix Mouse Gene 1.0 ST Array 10393848 - -
          Affymetrix Mouse Gene 1.0 ST Array 10393850 - -
          Affymetrix Mouse Gene 1.0 ST Array 10393846 - -
          Affymetrix Mouse Gene 1.0 ST Array 10393847 - -
          Affymetrix Mouse Gene 1.0 ST Array 10393845 - -
          Affymetrix HT MG-430 PM Array Plate 1436211_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1417724_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10393844 - -
          Mouse Genome 430 2.0 Array 1422993_PM_s_at - -
          Mouse Genome 430 2.0 Array 1417724_at - -
          Mouse Genome 430 2.0 Array 1436211_at - -
          Mouse Genome 430 2.0 Array 1417724_PM_at - -
          Mouse Genome 430 2.0 Array 1436211_PM_at - -
          Mouse Genome 430 2.0 Array 1422993_s_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 21681_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 21681_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1100040251.mm.1 TC1100004219.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100040247.mm.1 NM_011568.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100040247.mm.1 NM_011568.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100040247.mm.1 TC1100004219.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100040248.mm.1 NM_011568.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100040248.mm.1 NM_011568.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100040248.mm.1 TC1100004219.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100040249.mm.1 NM_011568.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100040249.mm.1 NM_011568.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100040249.mm.1 TC1100004219.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100040243.mm.1 NM_011568.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100040243.mm.1 NM_011568.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100040243.mm.1 TC1100004219.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100040250.mm.1 NM_011568.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100040250.mm.1 NM_011568.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100040250.mm.1 TC1100004219.mm.1 MTA TC
          Mouse Expression Set 430 1422993_s_at - -
          Mouse Expression Set 430 1417724_PM_at - -
          Mouse Expression Set 430 1422993_PM_s_at - -
          Mouse Expression Set 430 1417724_at - -
          Mouse Expression Set 430 1436211_at - -
          Mouse Expression Set 430 1436211_PM_at - -
          Murine Genome U74v2 Set 160633_PM_at - -
          Murine Genome U74v2 Set 160633_at - -
          Murine Genome U74v2 Set 135150_at - -
          Murine Genome U74v2 Set 135150_PM_at - -
          Mouse Genome 430A 2.0 Array 1422993_PM_s_at - -
          Mouse Genome 430A 2.0 Array 1422993_s_at - -
          Mouse Genome 430A 2.0 Array 1417724_at - -
          Mouse Genome 430A 2.0 Array 1417724_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1417724_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1417724_at - -
          Affymetrix HT Mouse Genome 430B Array 1436211_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1436211_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 871089 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 3800731 - -
          Illumina mouse-6 v1.0 expression beadchip 6400114 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003800731 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006400114 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2617245 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2747744 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2747744 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2617245 - -
          mouse-6 v1.1 (Illumina) scl0021681.1_61-S - -
          mouse-6 v1.1 (Illumina) scl021681.3_9-S - -
          Illumina Sentrix mouse-6.v1 bead array 3800731 - -
          Illumina Sentrix mouse-6.v1 bead array 6400114 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0021681.1_61-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl021681.3_9-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6400114 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3800731 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2747744 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2617245 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2747744 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2617245 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl021681.3_9-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl0021681.1_61-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-mrft7:80/100.66.76.145:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0