Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00488795_m1
          See other CLEC4A2 GE Assays ›
          Gene Symbol:
          Clec4a2
          Gene Name:
          C-type lectin domain family 4, member a2
          Gene Aliases:
          Clec4a, Clecsf6, DCIR, Dcir1
          Chromosome Location:
          Chr.6: 123122690 - 123143999 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00488795_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Clec4a2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001170332.1 NP_001163803.1 4 - 5 492 92
          NM_011999.4 NP_036129.1 3 - 4 572 92
          GenBank mRNA AF387099.1 - 3 - 4 577 92
          AJ133533.1 - 3 - 4 578 92
          AK049002.1 - 3 - 4 497 92
          BC075729.1 - 3 - 4 414 30287332 92
          BC096565.1 - 4 - 5 501 874396 92

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0600001401.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Clec4a2

          Panther Classification:

          Molecular Function -

          membrane traffic protein

          Gene Ontology Categories:

          Function(s) Process(es)

          immune system process
          innate immune response
          carbohydrate binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2039622 AK049002.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2039622 NM_001170333.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2039622 NM_011999.4 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2039622 NM_001170332.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 402518 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 272675 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 399484 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 689241 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 734292 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 756007 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 895071 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 271099 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 135613 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 153990 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P121565 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2039622 AK049002.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2039622 NM_001170333.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2039622 NM_011999.4 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2039622 NM_001170332.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P444092 BC075729.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P444092 BC096565.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P444092 AK049002.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P444092 AJ133533.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P444092 NM_011999.4 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24811531 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24811542 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24811528 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24811536 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24811533 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24811516 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24811534 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24811537 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24811539 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24811522 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24811524 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24811527 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24811523 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24811546 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24811547 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24811519 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24811526 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24811530 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24811540 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24811518 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P121565 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06053627 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06053647 - -
          Whole Mouse Genome G4122A A_52_P356106 AK049002.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P356106 NM_001170333.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P356106 NM_011999.4 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P356106 NM_001170332.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P353698 BC075729.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P353698 BC096565.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P353698 AK049002.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P353698 BC006623.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P353698 AJ133533.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P444092 BC075729.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P444092 BC096565.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P444092 AK049002.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P444092 AJ133533.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P444092 NM_011999.4 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P107724 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P140951 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P06053659 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P121565 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05367295 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05387956 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05387960 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05367296 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05367293 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05387962 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05387970 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05387968 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05387961 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05387969 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24811623 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24811647 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24811524 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24811642 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24811519 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24811628 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24811637 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24811644 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24811627 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24811557 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24811596 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24811630 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24811501 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24811570 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24811620 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24811631 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24811632 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24811536 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24811646 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24811605 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24811583 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24811581 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24811510 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24811556 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24811514 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24811597 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24811500 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24811531 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24811542 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24811615 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24811601 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24811558 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24811649 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24811505 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24811539 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24811604 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24811609 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24811561 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24811507 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24811639 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24811540 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24811614 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24811626 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24811502 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24811527 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24811530 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24811573 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24811511 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24811607 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24811533 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24811602 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24811618 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24811580 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24811537 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24811608 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24811599 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24811542 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24811507 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24811523 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24811554 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24811537 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24811505 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24811541 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24811556 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24811569 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05543234 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05025861 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05367296 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05655330 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24811531 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24811550 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24811511 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24811568 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24811530 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24811536 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24811558 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24811527 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24811557 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05647934 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24811510 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05367295 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24811533 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24811539 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24811516 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24811564 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05367293 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05655326 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24811514 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24811524 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05514837 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24811540 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10541595 - -
          Affymetrix Mouse Gene 1.0 ST Array 10541598 - -
          Affymetrix Mouse Gene 1.0 ST Array 10541592 - -
          Affymetrix Mouse Gene 1.0 ST Array 10541589 - -
          Affymetrix Mouse Gene 1.0 ST Array 10541591 - -
          Affymetrix Mouse Gene 1.0 ST Array 10541590 - -
          Affymetrix Mouse Gene 1.0 ST Array 10541588 - -
          Affymetrix Mouse Gene 1.0 ST Array 10541594 - -
          Affymetrix Mouse Gene 1.0 ST Array 10541596 - -
          Affymetrix Mouse Gene 1.0 ST Array 10541597 - -
          Affymetrix HT MG-430 PM Array Plate 1425406_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1422013_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10541587 - -
          Mouse Genome 430 2.0 Array 1425407_PM_s_at - -
          Mouse Genome 430 2.0 Array 1425406_PM_at - -
          Mouse Genome 430 2.0 Array 1425407_s_at - -
          Mouse Genome 430 2.0 Array 1425406_at - -
          Mouse Genome 430 2.0 Array 1422013_at - -
          Mouse Genome 430 2.0 Array 1422013_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 26888_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 26888_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0600011292.mm.1 TC0600001401.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600011292.mm.1 NM_001170333.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600011292.mm.1 NM_001170333.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600011292.mm.1 NM_011999.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600011292.mm.1 NM_011999.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0600011297.mm.1 TC0600001401.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600011297.mm.1 NM_001170332.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600011297.mm.1 NM_001170332.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600011297.mm.1 NM_001170333.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600011297.mm.1 NM_001170333.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600011297.mm.1 NM_011999.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600011297.mm.1 NM_011999.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0600011298.mm.1 TC0600001401.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600011298.mm.1 NM_001170332.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600011298.mm.1 NM_001170332.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600011298.mm.1 NM_001170333.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600011298.mm.1 NM_001170333.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600011298.mm.1 NM_011999.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600011298.mm.1 NM_011999.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0600011299.mm.1 TC0600001401.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600011299.mm.1 NM_001170332.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600011299.mm.1 NM_001170332.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600011299.mm.1 NM_001170333.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600011299.mm.1 NM_001170333.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600011299.mm.1 NM_011999.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600011299.mm.1 NM_011999.4 Refseq
          Mouse Expression Set 430 1425407_s_at - -
          Mouse Expression Set 430 1422013_PM_at - -
          Mouse Expression Set 430 1425406_PM_at - -
          Mouse Expression Set 430 1422013_at - -
          Mouse Expression Set 430 1425406_at - -
          Mouse Expression Set 430 1425407_PM_s_at - -
          Murine Genome U74v2 Set 92762_at - -
          Murine Genome U74v2 Set 92762_PM_at - -
          Mouse Genome 430A 2.0 Array 1425406_PM_at - -
          Mouse Genome 430A 2.0 Array 1422013_PM_at - -
          Mouse Genome 430A 2.0 Array 1425407_s_at - -
          Mouse Genome 430A 2.0 Array 1425407_PM_s_at - -
          Mouse Genome 430A 2.0 Array 1425406_at - -
          Mouse Genome 430A 2.0 Array 1422013_at - -
          Affymetrix HT Mouse Genome 430A Array 1422013_at - -
          Affymetrix HT Mouse Genome 430A Array 1425406_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1425406_at - -
          Affymetrix HT Mouse Genome 430A Array 1422013_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 922417 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 2470292 - -
          Illumina mouse-6 v1.0 expression beadchip 2470292 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002470292 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2739466 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2739466 - -
          mouse-6 v1.1 (Illumina) scl29530.8_166-S - -
          Illumina Sentrix mouse-6.v1 bead array 2470292 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl29530.8_166-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2470292 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2819015 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2739466 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2819015 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2739466 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline