Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00489055_m1
          See other CD52 GE Assays ›
          Gene Symbol:
          Cd52
          Gene Name:
          CD52 antigen
          Gene Aliases:
          AI463198, B7, B7-Ag, CAMPATH-1, CLS1, MB7
          Chromosome Location:
          Chr.4: 134093538 - 134095073 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00489055_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cd52
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_013706.2 NP_038734.1 1 - 2 143 81
          GenBank mRNA AK088161.1 - 1 - 2 110 81
          BC027495.1 - 1 - 2 93 1396480 81
          M55561.1 - 1 - 2 132 81
          X76696.1 - 1 - 2 119 81

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0400003724.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cd52

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23321062 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P151126 NM_013706.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P151126 AK088161.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P151126 AK155728.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P151126 BC027495.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P151126 M55561.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P151126 NM_013706.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P151126 AK088161.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P151126 AK155728.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P151126 BC027495.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P151126 M55561.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 547825 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P151126 NM_013706.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P151126 AK088161.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P151126 AK155728.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P151126 BC027495.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P151126 M55561.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P151126 NM_013706.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P151126 AK088161.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P151126 AK155728.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P151126 BC027495.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P151126 M55561.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23321018 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23321043 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23321007 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23321051 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23321044 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23321036 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23321011 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23321034 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23321021 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23321045 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23321016 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23321047 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23321062 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23321054 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23321052 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23321023 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23321025 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23321027 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23321015 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23321056 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23321040 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23321030 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23321037 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23321035 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23321010 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23321042 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23321028 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23321049 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23321059 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23321019 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23321014 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23321058 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23321032 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23321009 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23321020 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23321038 - -
          Whole Mouse Genome G4122A A_51_P151126 NM_013706.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P151126 AK088161.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P151126 AK155728.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P151126 BC027495.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P151126 M55561.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P151126 NM_013706.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P151126 AK088161.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P151126 AK155728.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P151126 BC027495.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P151126 M55561.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P109632 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03579971 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03579965 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03343540 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03579977 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03343542 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03579983 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03379483 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03343545 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23321062 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23320998 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23321028 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23320984 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23321051 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23321004 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23320981 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23321069 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23321020 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23320975 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23321015 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23320968 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23320999 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23320966 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23321058 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23321023 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23321042 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23320963 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23321044 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23321029 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23320988 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23321030 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23321009 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23321043 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23320974 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23320978 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23321005 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23321011 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23321037 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23320967 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23321049 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23320997 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23321066 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23321010 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23321018 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23320987 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23320996 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23321016 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23321014 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23321047 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23321007 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23321035 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23320993 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23320976 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23321062 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23320982 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23321054 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23321034 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23320972 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23321027 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23320971 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23321002 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23321040 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23321025 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23320989 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03579967 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23320981 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23321025 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03633329 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23320975 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03359960 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04018272 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04076535 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23320998 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03616801 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23321029 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23321034 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23320991 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03359962 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23321028 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23321023 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03579996 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03969859 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23321015 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04076539 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03359969 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03579976 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23321009 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23320963 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23321021 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23321019 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04076538 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03609168 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23321014 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03819500 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03969860 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03972882 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23320996 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03579991 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03237047 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03527754 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23320966 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03359968 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03237043 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03353848 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03579968 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23320989 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23321020 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03633327 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23320980 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23321027 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03359967 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04076542 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04090684 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23321011 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03633334 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04046478 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03579978 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23321010 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03237041 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03969862 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10517167 - -
          Affymetrix Mouse Gene 1.0 ST Array 10517166 - -
          Affymetrix Mouse Gene 1.0 ST Array 10517168 - -
          Affymetrix HT MG-430 PM Array Plate 1460218_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10517165 - -
          Mouse Genome 430 2.0 Array 1460218_PM_at - -
          Mouse Genome 430 2.0 Array 1460218_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 23833_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 23833_PM_at - -
          Affymetrix Murine Genome 6500A Array Msa10540_at - -
          Affymetrix Mouse Clariom S Array PSR0400030604.mm.1 TC0400003724.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400030604.mm.1 NM_013706.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400030604.mm.1 NM_013706.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0400030609.mm.1 TC0400003724.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400030609.mm.1 NM_013706.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400030609.mm.1 NM_013706.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0400030606.mm.1 TC0400003724.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400030606.mm.1 NM_013706.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400030606.mm.1 NM_013706.2 Refseq
          Mouse Expression Set 430 1460218_PM_at - -
          Mouse Expression Set 430 1460218_at - -
          Murine Genome U74v2 Set 104606_PM_at - -
          Murine Genome U74v2 Set 104606_at - -
          Mouse Genome 430A 2.0 Array 1460218_at - -
          Mouse Genome 430A 2.0 Array 1460218_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1460218_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1460218_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 926003 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 5910242 - -
          Illumina mouse-6 v1.0 expression beadchip 5910242 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005910242 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1251171 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1251171 - -
          mouse-6 v1.1 (Illumina) scl023833.2_2-S - -
          Illumina Sentrix mouse-6.v1 bead array 5910242 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl023833.2_2-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5910242 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2910934 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2910934 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2771380 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl023833.2_2-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-7b6576f7bc-knt4q:80/100.66.75.14:80.
          git-commit: 6d309ebe7a31b2ceadeb87f43b473eebab110cf3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-2026.03.11-1.0