| Array Name |
Probe IDs |
Targets |
Target Type |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P26343223 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P26343245 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P26343221 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P26343247 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P26343222 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P26343244 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P26343220 |
- |
- |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2034670 |
AK137282.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2034670 |
AK029429.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2034670 |
AK030910.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2034670 |
AK031262.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2034670 |
AF176525.1 |
mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
895669 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
365290 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
556228 |
AB039275.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
556228 |
BC132090.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
556228 |
BC132665.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
556228 |
NM_020268.3 |
refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
1117 |
- |
- |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2034670 |
AK137282.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2034670 |
AK029429.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2034670 |
AK030910.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2034670 |
AK031262.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2034670 |
AF176525.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P380541 |
BC054471.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P380541 |
AK137282.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P380541 |
BC005699.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P380541 |
AK029429.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P380541 |
AK030910.1 |
mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26343212 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26343214 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26343229 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26343245 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26343252 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26343216 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26343222 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26343227 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26343256 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26343224 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26343250 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26343220 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26343254 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26343247 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26343249 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26343217 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26343219 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26343251 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26343226 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P01880801 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P06716516 |
- |
- |
| Whole Mouse Genome G4122A |
A_51_P380541 |
BC054471.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P380541 |
AK137282.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P380541 |
BC005699.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P380541 |
AK029429.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P380541 |
AK030910.1 |
mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P113161 |
- |
- |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_67_P01880801 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P07827113 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P08065762 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P08065765 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P08149835 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P08065768 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P08000359 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P08149836 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P08065769 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P08149848 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P26343222 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P26343244 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P26343223 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P26343247 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P26343245 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P26343221 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P26343220 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26343224 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26343183 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26343196 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26343293 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26343189 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26343242 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26343255 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26343243 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26343273 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26343207 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26343245 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26343198 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26343239 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26343295 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26343226 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26343249 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26343253 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26343208 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26343270 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26343291 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26343197 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26343251 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26343200 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26343212 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26343221 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26343269 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26343285 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26343184 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26343222 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26343186 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26343271 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26343282 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26343250 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26343252 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26343216 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26343214 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26343220 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26343279 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26343192 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26343233 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26343247 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26343283 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26343229 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26343231 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26343203 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26343281 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26343187 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26343227 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26343254 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26343253 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P08149846 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26343214 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26343196 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26343222 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26343227 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26343245 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P07843312 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26343207 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26343190 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P07827113 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P07534890 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26343212 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P08149835 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26343198 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26343239 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26343203 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26343217 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P07956370 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P08007973 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P08065768 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26343221 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26343216 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26343247 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26343188 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26343243 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26343249 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P08187405 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P08007978 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P08086961 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26343195 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26343189 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26343192 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26343250 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26343218 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26343224 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26343208 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26343252 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26343219 |
- |
- |