Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00489467_s1
          See other MTAG2 GE Assays ›
          Gene Symbol:
          Mtag2
          Gene Name:
          metastasis associated gene 2
          Gene Aliases:
          MAG
          Chromosome Location:
          Chr.7: 45366163 - 45367948 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00489467_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Mtag2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_015480.1 2 323 54
          NR_027802.1 1 352 54
          GenBank mRNA AK042966.1 - 2 323 54
          BC107317.1 - 1 352 54
          BC107318.1 - 1 352 54
          BC119507.1 - 1 352 54
          BC119508.1 - 1 352 54
          BC127168.1 - 1 352 54
          BC127169.1 - 1 352 54
          U88401.1 - 1 350 54

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700000841.mm.1

          Set Membership:

          Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P511608 NR_027802.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P511608 AK042966.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P511608 U88401.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P511608 NR_027802.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P511608 AK042966.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P511608 U88401.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 172007 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P511608 NR_027802.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P511608 AK042966.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P511608 U88401.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P511608 NR_027802.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P511608 AK042966.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P511608 U88401.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P511608 NR_027802.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P511608 AK042966.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P511608 U88401.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P511608 NR_027802.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P511608 AK042966.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P511608 U88401.1 mouse_transcriptome
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10552930 - -
          Affymetrix Mouse Gene 1.0 ST Array 10552931 - -
          Affymetrix HT MG-430 PM Array Plate 1420605_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10563253 - -
          Affymetrix Mouse Gene 1.1 ST Array 10552929 - -
          Mouse Genome 430 2.0 Array 1420605_PM_at - -
          Mouse Genome 430 2.0 Array 1420605_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 50994_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 50994_at - -
          Affymetrix Mouse Clariom S Array PSR0700007246.mm.1 TC0700000841.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700007246.mm.1 NR_027802.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700007246.mm.1 NR_027802.1 Refseq
          Mouse Expression Set 430 1420605_at - -
          Mouse Expression Set 430 1420605_PM_at - -
          Murine Genome U74v2 Set 96505_at - -
          Murine Genome U74v2 Set 96505_PM_at - -
          Mouse Genome 430A 2.0 Array 1420605_at - -
          Mouse Genome 430A 2.0 Array 1420605_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1420605_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1420605_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 304701 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2692688 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2692688 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2692688 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline