Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00489940_m1
          See other CLPP GE Assays ›
          Gene Symbol:
          Clpp
          Gene Name:
          caseinolytic mitochondrial matrix peptidase proteolytic subunit
          Gene Aliases:
          AU019820, D17Wsu160e
          Chromosome Location:
          Chr.17: 56990264 - 56996371 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00489940_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Clpp
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_017393.2 NP_059089.1 3 - 4 417 63
          GenBank mRNA AJ005253.1 - 3 - 4 385 63
          AK002238.1 - 3 - 4 356 63
          AK004024.1 - 3 - 4 366 63
          AK145765.1 - 3 - 4 379 63
          AK147719.1 - 3 - 4 345 63
          AK168053.1 - 3 - 4 364 63
          BC001998.1 - 3 - 4 376 3487489 63

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1700001020.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Protease Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Clpp

          Panther Classification:

          Molecular Function -

          serine protease

          Gene Ontology Categories:

          Function(s) Process(es)

          proteolysis
          misfolded or incompletely synthesized protein catabolic process
          protein homooligomerization
          proteolysis involved in cellular protein catabolic process
          serine-type endopeptidase activity
          protein binding
          peptidase activity
          serine-type peptidase activity
          hydrolase activity
          identical protein binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31259432 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31259429 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31259430 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P134739 BC001998.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P134739 AK168053.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P134739 AK004024.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P134739 NM_017393.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P134739 AJ005253.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 488955 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 190626 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 271097 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 764637 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 124508 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P118674 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P134739 BC001998.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P134739 AK168053.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P134739 AK004024.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P134739 NM_017393.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P134739 AJ005253.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P139021 AJ005253.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P139021 BC001998.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P139021 AK168053.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P139021 AK004024.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P139021 NM_017393.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31259440 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31259435 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31259437 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31259438 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31259453 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31259430 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31259433 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31259434 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31259442 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31259431 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31259455 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31259443 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31259432 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31259458 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31259446 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31259459 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31259445 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31259460 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31259451 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31259448 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31259454 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P118674 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P03340627 - -
          Whole Mouse Genome G4122A A_51_P139021 AJ005253.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P139021 BC001998.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P139021 AK168053.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P139021 AK004024.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P139021 NM_017393.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P134739 BC001998.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P134739 AK168053.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P134739 AK004024.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P134739 NM_017393.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P134739 AJ005253.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P118674 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13434760 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31259432 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31259429 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31259430 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31259478 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31259459 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31259484 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31259472 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31259435 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31259492 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31259445 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31259391 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31259379 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31259486 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31259460 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31259520 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31259399 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31259481 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31259490 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31259482 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31259430 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31259433 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31259419 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31259446 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31259423 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31259390 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31259427 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31259382 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31259475 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31259487 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31259411 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31259468 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31259463 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31259424 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31259526 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31259499 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31259509 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31259529 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31259464 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31259518 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31259473 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31259521 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31259408 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31259386 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31259440 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31259497 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31259403 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31259515 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31259406 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31259513 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31259511 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31259443 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31259421 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31259458 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31259434 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31259525 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31259516 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31259432 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31259488 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31259507 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31259381 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31259494 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31259479 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31259393 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31259504 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31259438 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31259470 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31259466 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31259442 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31259496 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31259394 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31259451 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31259454 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31259477 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31259527 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31259409 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31259530 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31259505 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31259493 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31259398 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31259404 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31259437 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31259454 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31259424 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13540142 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31259438 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13294095 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31259415 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13434764 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31259431 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31259435 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13456132 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31259442 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31259446 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31259411 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13421979 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13162940 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31259425 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13200857 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31259421 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31259443 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13425818 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31259458 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31259430 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13456136 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31259460 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13407529 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31259448 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31259419 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31259427 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31259440 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31259459 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13423225 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13434752 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31259434 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10446208 - -
          Affymetrix Mouse Gene 1.0 ST Array 10446210 - -
          Affymetrix Mouse Gene 1.0 ST Array 10446211 - -
          Affymetrix Mouse Gene 1.0 ST Array 10446212 - -
          Affymetrix Mouse Gene 1.0 ST Array 10446209 - -
          Affymetrix Mouse Gene 1.0 ST Array 10446213 - -
          Affymetrix HT MG-430 PM Array Plate 1416615_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1416616_PM_s_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10446207 - -
          Mouse Genome 430 2.0 Array 1416615_PM_at - -
          Mouse Genome 430 2.0 Array 1416616_PM_s_at - -
          Mouse Genome 430 2.0 Array 1416615_at - -
          Mouse Genome 430 2.0 Array 1416616_s_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 53895_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 53895_at - -
          Affymetrix Mouse Clariom S Array PSR1700009841.mm.1 NM_017393.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700009841.mm.1 NM_017393.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700009841.mm.1 TC1700001020.mm.1 MTA TC
          Mouse Expression Set 430 1416616_s_at - -
          Mouse Expression Set 430 1416616_PM_s_at - -
          Mouse Expression Set 430 1416615_PM_at - -
          Mouse Expression Set 430 1416615_at - -
          Murine Genome U74v2 Set 93048_PM_at - -
          Murine Genome U74v2 Set 93048_at - -
          Mouse Genome 430A 2.0 Array 1416616_s_at - -
          Mouse Genome 430A 2.0 Array 1416615_PM_at - -
          Mouse Genome 430A 2.0 Array 1416615_at - -
          Mouse Genome 430A 2.0 Array 1416616_PM_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1416616_PM_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1416616_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1416615_at - -
          Affymetrix HT Mouse Genome 430A Array 1416615_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 406804 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 6520102 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006520102 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2771009 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2771009 - -
          mouse-6 v1.1 (Illumina) scl50582.4.1_9-S - -
          Illumina Sentrix mouse-6.v1 bead array 6520102 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl50582.4.1_9-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6520102 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2982890 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2982890 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2771009 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl50582.4.1_9-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-4lm45:80/100.66.75.98:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0