Search
Search

Crtam| Gene Symbol: Crtam | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001281954.1 | NP_001268883.1 | 8 - 9 | 1012 | 85 | |
| NM_019465.4 | NP_062338.3 | 8 - 9 | 1009 | 85 | ||
| GenBank mRNA | AF001104.1 | - | 8 - 9 | 955 | 85 | |
| AK163744.1 | - | 3 - 4 | 410 | 85 | ||
| BC117715.1 | - | 8 - 9 | 957 | 85 | ||
| BC117716.1 | - | 8 - 9 | 958 | 40088099 | 85 | |
Set Membership: |
|