Search
Search

Stub1| Gene Symbol: Stub1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019719.3 | NP_062693.1 | 1 - 2 | 673 | 94 | |
| GenBank mRNA | AF129086.1 | - | 1 - 2 | 220 | 94 | |
| AK002752.1 | - | 1 - 2 | 230 | 94 | ||
| AK004464.1 | - | 1 - 2 | 221 | 94 | ||
| AK045776.1 | - | 1 - 2 | 222 | 94 | ||
| AK158595.1 | - | 1 - 2 | 257 | 94 | ||
| AK166630.1 | - | 1 - 2 | 239 | 94 | ||
| BC020110.1 | - | 1 - 2 | 215 | 94 | ||
| BC027427.1 | - | 1 - 2 | 673 | 4191866 | 94 | |
| BC038939.1 | - | 1 - 2 | 262 | 2598940 | 94 | |
Set Membership: |
|