Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00490735_m1
          See other CYP11A1 GE Assays ›
          Gene Symbol:
          Cyp11a1
          Gene Name:
          cytochrome P450, family 11, subfamily a, polypeptide 1
          Gene Aliases:
          Cyp11a, Cypxia1, D9Ertd411e, P450scc, Scc, cscc
          Chromosome Location:
          Chr.9: 58015017 - 58027023 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00490735_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information
          • Citations (1)

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cyp11a1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_019779.3 NP_062753.3 5 - 6 1002 82
          GenBank mRNA AF195119.1 - 5 - 6 1002 82
          BC068264.1 - 5 - 6 1021 30014890 82

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0900000722.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection P450 Recommended Assay Mus musculus Mouse Androgen Panel 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cyp11a1

          Gene Ontology Categories:

          Function(s) Process(es)

          lipid metabolic process
          steroid biosynthetic process
          C21-steroid hormone biosynthetic process
          mating behavior
          steroid metabolic process
          cholesterol metabolic process
          response to hydrogen peroxide
          oxidation-reduction process
          monooxygenase activity
          iron ion binding
          cholesterol monooxygenase (side-chain-cleaving) activity
          cholesterol binding
          oxidoreductase activity
          oxidoreductase activity, acting on paired donors, with incorporation or reduction of molecular oxygen
          heme binding
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P309733 BC068264.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P309733 NM_019779.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P309733 AF195119.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P309733 BC068264.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P309733 NM_019779.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P309733 AF195119.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 941897 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 910676 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 734780 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 789515 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 853048 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 700142 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 654798 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 535095 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 127688 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 548512 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 122962 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 34470 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P165635 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P136981 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P309733 BC068264.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P309733 NM_019779.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P309733 AF195119.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P309733 BC068264.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P309733 NM_019779.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P309733 AF195119.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P309727 BC068264.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P309727 NM_019779.3 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P309727 AF195119.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P309733 BC068264.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P309733 NM_019779.3 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P309733 AF195119.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P309733 BC068264.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P309733 NM_019779.3 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P309733 AF195119.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06803194 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01942670 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P136981 - -
          Whole Mouse Genome G4122A A_52_P475697 AK077945.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P475697 AK135608.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P309727 BC068264.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P309727 NM_019779.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P309727 AF195119.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P309733 BC068264.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P309733 NM_019779.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P309733 AF195119.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P309733 BC068264.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P309733 NM_019779.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P309733 AF195119.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P114673 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P104399 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P132526 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P136981 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P06803135 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07824751 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07824752 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07824755 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07824758 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07824754 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26545159 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26545171 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26545165 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26545160 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26545164 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26545176 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26545161 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26545167 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26545157 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26545173 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26545170 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26545169 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07871596 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26545157 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07755964 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26544975 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26545003 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07611510 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26545169 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07611509 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26545164 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26544962 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07727482 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26544970 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26545154 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26544998 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26544977 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07611508 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26544992 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26544960 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26544961 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07702977 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26544939 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26545159 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26545027 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07722770 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26544989 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26545160 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26544958 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26544996 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07702972 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08113173 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26544983 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07624097 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26545005 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26544956 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26545010 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26544946 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07702959 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26545007 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26545020 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08051639 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07725388 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08051638 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08051633 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26544985 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26544967 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07535443 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26544986 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26545167 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07722769 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26544964 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26545165 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26544966 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08051619 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26545012 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26544969 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07702978 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26544987 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08051648 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08051630 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26545000 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26545022 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26545033 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07846083 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26545029 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26544981 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07727478 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26545009 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07607577 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26544973 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07541263 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26544940 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26544965 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26545032 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26545011 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26545004 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07702966 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10585801 - -
          Affymetrix Mouse Gene 1.0 ST Array 10585799 - -
          Affymetrix Mouse Gene 1.0 ST Array 10585798 - -
          Affymetrix Mouse Gene 1.0 ST Array 10585797 - -
          Affymetrix Mouse Gene 1.0 ST Array 10585795 - -
          Affymetrix Mouse Gene 1.0 ST Array 10585796 - -
          Affymetrix Mouse Gene 1.0 ST Array 10585800 - -
          Affymetrix Mouse Gene 1.0 ST Array 10585802 - -
          Affymetrix HT MG-430 PM Array Plate 1457604_PM_x_at - -
          Affymetrix HT MG-430 PM Array Plate 1448804_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1439947_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10585794 - -
          Mouse Genome 430 2.0 Array 1439947_PM_at - -
          Mouse Genome 430 2.0 Array 1448804_at - -
          Mouse Genome 430 2.0 Array 1439947_at - -
          Mouse Genome 430 2.0 Array 1457604_PM_x_at - -
          Mouse Genome 430 2.0 Array 1448804_PM_at - -
          Mouse Genome 430 2.0 Array 1457604_x_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 13070_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 13070_at - -
          Affymetrix Mouse Clariom S Array PSR0900005311.mm.1 NM_019779.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0900005311.mm.1 TC0900000722.mm.1 MTA TC
          Mouse Expression Set 430 1448804_PM_at - -
          Mouse Expression Set 430 1448804_at - -
          Mouse Genome 430A 2.0 Array 1448804_at - -
          Mouse Genome 430A 2.0 Array 1448804_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1448804_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1448804_at - -
          Affymetrix HT Mouse Genome 430B Array 1439947_at - -
          Affymetrix HT Mouse Genome 430B Array 1457604_PM_x_at - -
          Affymetrix HT Mouse Genome 430B Array 1457604_x_at - -
          Affymetrix HT Mouse Genome 430B Array 1439947_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 646473 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 101400397 - -
          MouseWG-6 v2 Expression BeadChip 106590086 - -
          MouseWG-6 v2 Expression BeadChip 3450037 - -
          Illumina mouse-6 v1.0 expression beadchip 101400397 - -
          Illumina mouse-6 v1.0 expression beadchip 3450037 - -
          Illumina mouse-6 v1.0 expression beadchip 106590086 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003450037 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2634367 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1242228 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2634367 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2467151 - -
          mouse-6 v1.1 (Illumina) scl36891.2_185-S - -
          mouse-6 v1.1 (Illumina) scl0013070.2_102-S - -
          Illumina Sentrix mouse-6.v1 bead array 3450037 - -
          Illumina Sentrix mouse-6.v1 bead array 106590086 - -
          Illumina Sentrix mouse-6.v1 bead array 101400397 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0013070.2_102-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3450037 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 101400397 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 106590086 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2634367 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2634367 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2467151 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2581647 - -

          Back To Top
           

          Citations (1)

          Effects of gestational diethylstilbestrol treatment on male and female gonads during early embryonic development.
          Authors
          Ikeda Y, Tanaka H, Esaki M
          Journal
          Endocrinology - Page No 3970 (Fri Aug 01 00:00:00 EDT 2008)
          Volume (Year)
          149 (2008)
          Product Usage
          -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline