Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00490782_m1
          See other CTSF GE Assays ›
          Gene Symbol:
          Ctsf
          Gene Name:
          cathepsin F
          Gene Aliases:
          AI481912
          Chromosome Location:
          Chr.19: 4855129 - 4860912 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00490782_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ctsf
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_019861.1 NP_063914.1 5 - 6 685 99
          GenBank mRNA AF136280.1 - 5 - 6 586 99
          AF197480.1 - 5 - 6 630 99
          AJ131851.1 - 5 - 6 562 99
          AK017474.1 - 5 - 6 571 99
          AK075862.1 - 5 - 6 578 99
          BC004054.1 - 2 - 3 156 99
          BC051131.1 - 4 - 5 804 99
          BC058758.1 - 5 - 6 576 6492834 99

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1900000046.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Protease Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ctsf

          Panther Classification:

          Molecular Function -

          cysteine protease

          Gene Ontology Categories:

          Function(s) Process(es)

          proteolysis
          proteolysis involved in cellular protein catabolic process
          cysteine-type endopeptidase activity
          peptidase activity
          cysteine-type peptidase activity
          hydrolase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31925432 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31925435 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31925433 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P660945 BC058758.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P660945 BC004054.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P660945 BC051131.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P660945 AK017474.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P660945 AJ131851.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P660945 BC058758.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P660945 BC004054.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P660945 BC051131.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P660945 AK017474.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P660945 AJ131851.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 321648 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 700969 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 283566 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 189523 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 64443 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P131777 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P660945 BC058758.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P660945 BC004054.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P660945 BC051131.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P660945 AK017474.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P660945 AJ131851.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P660945 BC058758.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P660945 BC004054.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P660945 BC051131.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P660945 AK017474.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P660945 AJ131851.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P660945 BC058758.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P660945 BC004054.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P660945 BC051131.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P660945 AK017474.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P660945 AJ131851.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P660945 BC058758.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P660945 BC004054.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P660945 BC051131.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P660945 AK017474.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P660945 AJ131851.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31925422 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31925445 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31925443 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31925430 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31925438 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31925433 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31925447 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31925442 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31925436 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31925437 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31925424 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31925441 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31925446 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31925432 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31925440 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31925427 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P131777 - -
          Whole Mouse Genome G4122A A_52_P660945 BC058758.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P660945 BC004054.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P660945 BC051131.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P660945 AK017474.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P660945 AJ131851.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P660945 BC058758.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P660945 BC004054.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P660945 BC051131.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P660945 AK017474.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P660945 AJ131851.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P100514 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P100166 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P131777 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14192644 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14399632 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14405005 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31925432 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31925433 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31925435 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31925517 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31925433 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31925521 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31925529 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31925420 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31925540 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31925442 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31925461 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31925474 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31925531 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31925537 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31925430 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31925432 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31925463 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31925412 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31925427 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31925424 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31925417 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31925407 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31925451 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31925526 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31925455 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31925527 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31925440 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31925441 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31925449 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31925452 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31925403 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31925469 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31925446 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31925534 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31925525 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31925464 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31925456 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31925518 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31925413 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31925443 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31925467 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31925437 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31925471 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31925458 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31925401 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31925438 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31925445 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14425503 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14320456 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31925457 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31925467 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31925441 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14334601 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14192654 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31925452 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31925422 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14373385 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31925472 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31925409 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14456648 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14381260 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31925440 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14192644 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31925442 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31925407 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31925437 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14192650 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31925430 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14334602 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14373378 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14345329 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31925421 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31925454 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14373388 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31925424 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31925419 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31925455 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31925471 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31925404 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31925428 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31925456 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14380038 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31925427 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31925401 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31925451 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14292230 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31925474 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31925464 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31925458 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31925403 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10460470 - -
          Affymetrix Mouse Gene 1.0 ST Array 10460479 - -
          Affymetrix Mouse Gene 1.0 ST Array 10460483 - -
          Affymetrix Mouse Gene 1.0 ST Array 10460469 - -
          Affymetrix Mouse Gene 1.0 ST Array 10460484 - -
          Affymetrix Mouse Gene 1.0 ST Array 10460475 - -
          Affymetrix Mouse Gene 1.0 ST Array 10460472 - -
          Affymetrix Mouse Gene 1.0 ST Array 10460478 - -
          Affymetrix Mouse Gene 1.0 ST Array 10460473 - -
          Affymetrix Mouse Gene 1.0 ST Array 10460476 - -
          Affymetrix Mouse Gene 1.0 ST Array 10460480 - -
          Affymetrix Mouse Gene 1.0 ST Array 10460477 - -
          Affymetrix Mouse Gene 1.0 ST Array 10460482 - -
          Affymetrix Mouse Gene 1.0 ST Array 10460474 - -
          Affymetrix Mouse Gene 1.0 ST Array 10460471 - -
          Affymetrix Mouse Gene 1.0 ST Array 10460481 - -
          Affymetrix HT MG-430 PM Array Plate 1451019_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10460468 - -
          Mouse Genome 430 2.0 Array 1451019_at - -
          Mouse Genome 430 2.0 Array 1451019_PM_at - -
          Mouse Genome 430 2.0 Array 1449794_x_at - -
          Mouse Genome 430 2.0 Array 1449794_PM_x_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 56464_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 56464_at - -
          Affymetrix Mouse Clariom S Array PSR1900000643.mm.1 NM_019861.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900000643.mm.1 NM_019861.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900000643.mm.1 TC1900000046.mm.1 MTA TC
          Mouse Expression Set 430 1451019_PM_at - -
          Mouse Expression Set 430 1451019_at - -
          Murine Genome U74v2 Set 97336_PM_at - -
          Murine Genome U74v2 Set 97336_at - -
          Mouse Genome 430A 2.0 Array 1451019_at - -
          Mouse Genome 430A 2.0 Array 1451019_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1451019_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1451019_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 411121 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 6100021 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006100021 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2615879 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2615879 - -
          mouse-6 v1.1 (Illumina) scl52823.10.1_53-S - -
          Illumina Sentrix mouse-6.v1 bead array 6100021 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl52823.10.1_53-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6100021 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2615879 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2615879 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl52823.10.1_53-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline