Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00490816_g1
          See other CRIPT GE Assays ›
          Gene Symbol:
          Cript
          Gene Name:
          cysteine-rich PDZ-binding protein
          Gene Aliases:
          1200020A08Rik, AA589469
          Chromosome Location:
          Chr.17: 87025561 - 87035808 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00490816_g1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cript
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_019936.3 NP_064320.1 4 - 5 345 105
          GenBank mRNA AF151637.1 - 4 - 5 310 105
          AK004828.1 - 4 - 5 105
          AK008523.1 - 4 - 5 330 105
          AK029409.1 - 4 - 5 333 105
          AK213321.1 - 2 - 3 177 105
          BC002258.1 - 3 - 4 227 105
          BC023013.1 - 4 - 5 319 5369938 105

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1700001308.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cript

          Gene Ontology Categories:

          Function(s) Process(es)

          cytoplasmic microtubule organization
          protein localization to microtubule
          establishment of protein localization
          regulation of postsynaptic density protein 95 clustering
          microtubule binding
          PDZ domain binding
          protein complex binding
          scaffold protein binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31420724 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P353392 AF151637.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P353392 AF151637.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 715056 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 313277 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 825124 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 375212 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 768537 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 387098 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 149974 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P132490 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P114886 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P353392 AF151637.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P353392 AF151637.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P353392 AF151637.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P353392 AF151637.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31420738 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31420727 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31420742 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31420728 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31420740 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31420737 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31420743 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31420745 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31420732 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31420733 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31420730 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31420736 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31420726 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31420739 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P114886 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P132490 - -
          Whole Mouse Genome G4122A A_51_P353392 AF151637.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P353392 AF151637.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P649161 AK008523.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P649161 AK029409.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P649161 AK213321.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P649161 BC002258.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P649161 BC023013.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P130474 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P132490 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P114886 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13209611 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13209610 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13209609 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31420724 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31420739 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31420786 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31420836 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31420846 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31420823 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31420835 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31420791 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31420749 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31420763 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31420730 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31420723 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31420833 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31420850 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31420785 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31420838 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31420818 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31420726 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31420769 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31420740 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31420756 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31420820 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31420751 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31420830 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31420814 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31420798 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31420736 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31420840 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31420809 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31420808 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31420774 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31420821 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31420765 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31420816 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31420747 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31420829 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31420738 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31420777 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31420779 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31420801 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31420797 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31420729 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31420733 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31420788 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31420737 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31420742 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31420793 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31420804 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31420767 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31420827 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31420806 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31420828 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31420728 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31420781 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31420758 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31420802 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31420848 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31420737 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31420746 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31420783 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31420775 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31420820 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13478445 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31420756 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31420758 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13317142 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31420827 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31420785 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31420729 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31420779 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13141801 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13306578 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31420728 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31420774 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13201525 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13344825 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31420733 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31420754 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31420747 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31420726 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31420828 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31420738 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31420810 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31420821 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13209625 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31420739 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13490476 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31420781 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31420730 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31420829 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31420755 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31420751 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13340111 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31420753 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31420743 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31420777 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31420742 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31420736 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31420816 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13088085 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31420823 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31420814 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13280914 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31420748 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10447351 - -
          Affymetrix Mouse Gene 1.0 ST Array 10447352 - -
          Affymetrix Mouse Gene 1.0 ST Array 10447350 - -
          Affymetrix Mouse Gene 1.0 ST Array 10447353 - -
          Affymetrix HT MG-430 PM Array Plate 1423486_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1423487_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10447349 - -
          Mouse Genome 430 2.0 Array 1423487_at - -
          Mouse Genome 430 2.0 Array 1423487_PM_at - -
          Mouse Genome 430 2.0 Array 1423486_at - -
          Mouse Genome 430 2.0 Array 1423486_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 56724_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 56724_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1700011900.mm.1 NM_019936.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700011900.mm.1 NM_019936.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1700011900.mm.1 TC1700001308.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700011899.mm.1 NM_019936.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700011899.mm.1 NM_019936.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1700011899.mm.1 TC1700001308.mm.1 MTA TC
          Mouse Expression Set 430 1423486_at - -
          Mouse Expression Set 430 1423487_at - -
          Mouse Expression Set 430 1423487_PM_at - -
          Mouse Expression Set 430 1423486_PM_at - -
          Murine Genome U74v2 Set 93760_PM_at - -
          Murine Genome U74v2 Set 93760_at - -
          Mouse Genome 430A 2.0 Array 1423487_PM_at - -
          Mouse Genome 430A 2.0 Array 1423486_PM_at - -
          Mouse Genome 430A 2.0 Array 1423486_at - -
          Mouse Genome 430A 2.0 Array 1423487_at - -
          Affymetrix HT Mouse Genome 430A Array 1423486_at - -
          Affymetrix HT Mouse Genome 430A Array 1423486_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1423487_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1423487_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 923398 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 5390056 - -
          Illumina mouse-6 v1.0 expression beadchip 5390056 - -
          Illumina mouse-6 v1.0 expression beadchip 5390403 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005390403 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005390056 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2716730 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1223929 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1223929 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2716730 - -
          mouse-6 v1.1 (Illumina) scl056724.5_33-S - -
          mouse-6 v1.1 (Illumina) scl0056724.1_273-S - -
          Illumina Sentrix mouse-6.v1 bead array 5390056 - -
          Illumina Sentrix mouse-6.v1 bead array 5390403 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl056724.5_33-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0056724.1_273-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5390403 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5390056 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1223929 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2846373 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2846373 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1223929 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl056724.5_33-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline