Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00490934_m1
          See other CLEC4N GE Assays ›
          Gene Symbol:
          Clec4n
          Gene Name:
          C-type lectin domain family 4, member n
          Gene Aliases:
          Clec6a, Clecsf10, Nkcl
          Chromosome Location:
          Chr.6: 123229843 - 123247024 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00490934_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Clec4n
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001190320.1 NP_001177249.1 4 - 5 567 87
          NM_001190321.1 NP_001177250.1 4 - 5 555 87
          NM_020001.2 NP_064385.1 5 - 6 657 87
          GenBank mRNA AF240357.1 - 5 - 6 626 87
          AF240358.1 - 4 - 5 524 87
          AK131697.1 - 5 - 6 644 87
          AK154673.1 - 4 - 5 554 87
          AK171046.1 - 6 - 7 827 87
          BC023008.1 - 5 - 6 626 5376247 87

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0600001403.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Mus musculus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Clec4n

          Panther Classification:

          Molecular Function -

          membrane traffic protein

          Gene Ontology Categories:

          Function(s) Process(es)

          immune system process
          positive regulation of I-kappaB kinase/NF-kappaB signaling
          innate immune response
          positive regulation of cytokine secretion
          defense response to fungus
          protein binding
          carbohydrate binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2035018 BC023008.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2035018 AK089166.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2035018 AK131697.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2035018 AK154673.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2035018 AK171046.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 620909 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 189507 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 160672 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 102607 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 511721 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 750234 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 882874 AK042927.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 72528 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 148062 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P165723 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P110977 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2035018 BC023008.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2035018 AK089166.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2035018 AK131697.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2035018 AK154673.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2035018 AK171046.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P263471 BC023008.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P263471 AK131697.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P263471 AK154673.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P263471 AK171046.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P263471 NM_001190320.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24811833 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24811813 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24811837 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24811834 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24811832 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24811808 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24811803 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24811822 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24811838 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24811825 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24811819 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24811804 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24811827 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24811801 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24811811 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24811836 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24811809 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24811799 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24811826 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24811798 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24811821 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24811817 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24811806 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24811840 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24811805 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24811824 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24811842 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24811815 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24811807 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24811843 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06053789 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06053777 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P110977 - -
          Whole Mouse Genome G4122A A_51_P263471 BC023008.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P263471 AK131697.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P263471 AK154673.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P263471 AK171046.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P263471 NM_001190320.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P135245 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P123017 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P110977 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05422473 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05735184 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05422477 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05422480 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05119341 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24811819 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24811862 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24811830 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24811858 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24811890 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24811846 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24811898 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24811842 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24811877 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24811833 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24811832 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24811859 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24811825 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24811866 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24811826 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24811813 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24811834 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24811900 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24811899 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24811798 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24811851 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24811853 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24811844 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24811807 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24811838 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24811874 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24811860 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24811827 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24811852 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24811803 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24811821 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24811843 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24811840 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24811891 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24811824 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24811848 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24811801 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24811822 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24811809 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24811849 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24811868 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24811903 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24811894 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24811856 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24811836 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24811896 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24811811 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24811808 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24811815 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24811822 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24811806 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24811862 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24811803 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24811838 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05735177 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24811809 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05793492 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05113394 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05710352 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24811824 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05735178 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24811863 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24811801 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24811844 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05793494 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24811846 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05422477 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05735185 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24811819 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24811821 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24811851 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24811849 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24811832 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24811842 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24811830 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24811827 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24811837 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05232357 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24811826 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24811850 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24811813 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05735176 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24811848 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24811852 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24811853 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24811817 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24811856 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05710350 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24811840 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24811833 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24811799 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10541612 - -
          Affymetrix Mouse Gene 1.0 ST Array 10541607 - -
          Affymetrix Mouse Gene 1.0 ST Array 10541611 - -
          Affymetrix Mouse Gene 1.0 ST Array 10541610 - -
          Affymetrix Mouse Gene 1.0 ST Array 10541609 - -
          Affymetrix Mouse Gene 1.0 ST Array 10541613 - -
          Affymetrix Mouse Gene 1.0 ST Array 10541608 - -
          Affymetrix Mouse Gene 1.0 ST Array 10541606 - -
          Affymetrix HT MG-430 PM Array Plate 1419627_PM_s_at - -
          Affymetrix HT MG-430 PM Array Plate 1425951_PM_a_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10541605 - -
          Mouse Genome 430 2.0 Array 1419627_s_at - -
          Mouse Genome 430 2.0 Array 1425951_a_at - -
          Mouse Genome 430 2.0 Array 1419627_PM_s_at - -
          Mouse Genome 430 2.0 Array 1425951_PM_a_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 56620_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 56620_at - -
          Affymetrix Mouse Clariom S Array PSR0600011328.mm.1 TC0600001403.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600011328.mm.1 NM_001190320.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600011328.mm.1 NM_001190320.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600011328.mm.1 NM_001190321.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600011328.mm.1 NM_001190321.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600011328.mm.1 NM_020001.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600011328.mm.1 NM_020001.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0600011329.mm.1 TC0600001403.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600011329.mm.1 NM_001190320.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600011329.mm.1 NM_001190320.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600011329.mm.1 NM_001190321.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600011329.mm.1 NM_001190321.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600011329.mm.1 NM_020001.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600011329.mm.1 NM_020001.2 Refseq
          Mouse Expression Set 430 1425951_a_at - -
          Mouse Expression Set 430 1419627_PM_s_at - -
          Mouse Expression Set 430 1419627_s_at - -
          Mouse Expression Set 430 1425951_PM_a_at - -
          Mouse Genome 430A 2.0 Array 1419627_PM_s_at - -
          Mouse Genome 430A 2.0 Array 1425951_PM_a_at - -
          Mouse Genome 430A 2.0 Array 1425951_a_at - -
          Mouse Genome 430A 2.0 Array 1419627_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1419627_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1419627_PM_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1425951_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1425951_a_at - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 6550086 - -
          Illumina mouse-6 v1.0 expression beadchip 6550086 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006550086 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1216664 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1216664 - -
          mouse-6 v1.1 (Illumina) scl056620.6_30-S - -
          Illumina Sentrix mouse-6.v1 bead array 6550086 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl056620.6_30-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6550086 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2993314 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2993314 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl056620.6_30-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline