Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00491029_g1
          See other SSSCA1 GE Assays ›
          Gene Symbol:
          Sssca1
          Gene Name:
          Sjogren's syndrome/scleroderma autoantigen 1 homolog (human)
          Gene Aliases:
          1500016H19Rik, C184L
          Chromosome Location:
          Chr.19: 5730306 - 5731732 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00491029_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Sssca1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_020491.5 NP_065237.3 2 - 3 213 95
          GenBank mRNA AB012865.1 - 2 - 3 181 95
          AK005265.1 - 2 - 3 185 95
          AK011957.1 - 2 - 3 203 95
          AK012592.1 - 2 - 3 191 95
          AK012704.1 - 2 - 3 191 95
          BC021593.1 - 2 - 3 183 5102521 95
          BY081504.1 - 2 - 3 271 95

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1900000981.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Sssca1

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          protein binding
          identical protein binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31930557 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31930506 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31930503 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31930504 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31930505 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P287052 AK012592.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P287052 NM_020491.4 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P287052 AB012865.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P287052 AK005265.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P287052 AK011957.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P287052 AK012592.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P287052 NM_020491.4 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P287052 AB012865.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P287052 AK005265.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P287052 AK011957.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 401958 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P287052 AK012592.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P287052 NM_020491.4 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P287052 AB012865.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P287052 AK005265.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P287052 AK011957.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P287052 AK012592.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P287052 NM_020491.4 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P287052 AB012865.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P287052 AK005265.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P287052 AK011957.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31930553 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31930554 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31930494 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31930556 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31930548 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31930498 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31930501 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31930499 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31930543 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31930537 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31930511 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31930540 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31930502 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31930504 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31930497 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31930555 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31930538 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31930539 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31930557 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31930505 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31930508 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31930541 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31930507 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31930552 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31930547 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31930544 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31930535 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31930509 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31930551 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31930500 - -
          Whole Mouse Genome G4122A A_52_P287052 AK012592.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P287052 NM_020491.4 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P287052 AB012865.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P287052 AK005265.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P287052 AK011957.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P287052 AK012592.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P287052 NM_020491.4 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P287052 AB012865.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P287052 AK005265.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P287052 AK011957.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P140708 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P120156 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14251882 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14251863 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14251889 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14251904 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14251919 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14251920 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31930506 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31930503 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31930557 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31930505 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31930504 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31930540 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31930463 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31930428 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31930554 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31930548 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31930422 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31930467 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31930544 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31930457 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31930483 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31930430 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31930552 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31930470 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31930452 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31930421 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31930444 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31930486 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31930419 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31930534 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31930418 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31930507 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31930508 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31930497 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31930557 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31930417 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31930442 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31930541 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31930434 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31930537 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31930466 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31930549 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31930426 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31930473 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31930499 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31930490 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31930547 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31930481 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31930543 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31930445 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31930455 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31930555 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31930494 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31930502 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31930440 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31930538 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31930461 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31930448 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31930489 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31930539 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31930439 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31930500 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31930498 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31930437 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31930505 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31930432 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31930431 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31930454 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31930553 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31930509 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31930443 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31930556 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31930416 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31930423 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31930472 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31930535 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31930502 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31930509 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31930507 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31930505 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31930511 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31930499 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14251896 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31930500 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31930498 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31930508 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10465222 - -
          Affymetrix Mouse Gene 1.0 ST Array 10465223 - -
          Affymetrix Mouse Gene 1.0 ST Array 10465221 - -
          Affymetrix Mouse Gene 1.0 ST Array 10465219 - -
          Affymetrix HT MG-430 PM Array Plate 1417725_PM_a_at - -
          Affymetrix HT MG-430 PM Array Plate 1417726_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10465220 - -
          Affymetrix Mouse Gene 1.1 ST Array 10465215 - -
          Mouse Genome 430 2.0 Array 1417726_PM_at - -
          Mouse Genome 430 2.0 Array 1417725_PM_a_at - -
          Mouse Genome 430 2.0 Array 1417725_a_at - -
          Mouse Genome 430 2.0 Array 1417726_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 56390_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 56390_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1900008852.mm.1 NM_020491.5 Refseq
          Affymetrix Mouse Clariom S Array PSR1900008852.mm.1 TC1900000981.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900008855.mm.1 NM_020491.5 Refseq
          Affymetrix Mouse Clariom S Array PSR1900008855.mm.1 TC1900000981.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900008856.mm.1 NM_020491.5 Refseq
          Affymetrix Mouse Clariom S Array PSR1900008856.mm.1 TC1900000981.mm.1 MTA TC
          Mouse Expression Set 430 1417726_at - -
          Mouse Expression Set 430 1417725_a_at - -
          Mouse Expression Set 430 1417726_PM_at - -
          Mouse Expression Set 430 1417725_PM_a_at - -
          Mouse Genome 430A 2.0 Array 1417725_a_at - -
          Mouse Genome 430A 2.0 Array 1417726_at - -
          Mouse Genome 430A 2.0 Array 1417726_PM_at - -
          Mouse Genome 430A 2.0 Array 1417725_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1417726_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1417725_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1417726_at - -
          Affymetrix HT Mouse Genome 430A Array 1417725_a_at - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 1240402 - -
          Illumina mouse-6 v1.0 expression beadchip 1240402 - -
          Illumina mouse-6 v1.0 expression beadchip 780309 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000780309 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001240402 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2691493 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2691493 - -
          mouse-6 v1.1 (Illumina) scl056390.4_41-S - -
          mouse-6 v1.1 (Illumina) scl0056390.1_292-S - -
          Illumina Sentrix mouse-6.v1 bead array 1240402 - -
          Illumina Sentrix mouse-6.v1 bead array 780309 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl056390.4_41-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0056390.1_292-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 780309 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1240402 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2691493 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2691492 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2691493 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2691492 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl056390.4_41-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl0056390.1_292-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline