Search
Search

Fbxo3| Gene Symbol: Fbxo3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020593.2 | NP_065618.1 | 4 - 5 | 540 | 63 | |
| NM_212433.1 | NP_997598.1 | 4 - 5 | 540 | 63 | ||
| GenBank mRNA | AF233226.1 | - | 4 - 5 | 479 | 63 | |
| AK004544.2 | - | 4 - 5 | 492 | 63 | ||
| AK009024.1 | - | 4 - 5 | 479 | 63 | ||
| AK019550.1 | - | 4 - 5 | 480 | 63 | ||
| AK028048.1 | - | 4 - 5 | 502 | 63 | ||
| AK031185.1 | - | 4 - 5 | 479 | 63 | ||
| AK031428.1 | - | 4 - 5 | 500 | 63 | ||
| AK049870.1 | - | 4 - 5 | 498 | 63 | ||
| AK050195.1 | - | 4 - 5 | 492 | 63 | ||
| AK085686.1 | - | 4 - 5 | 495 | 63 | ||
| AK150097.1 | - | 4 - 5 | 492 | 63 | ||
| AK151130.1 | - | 4 - 5 | 491 | 63 | ||
| AK151857.1 | - | 4 - 5 | 491 | 63 | ||
| AK152780.1 | - | 4 - 5 | 491 | 63 | ||
| AK167075.1 | - | 4 - 5 | 478 | 63 | ||
| BC010212.1 | - | 4 - 5 | 441 | 63 | ||
| BC050758.1 | - | 4 - 5 | 493 | 63 | ||
| BC058253.1 | - | 4 - 5 | 497 | 6465107 | 63 | |
Set Membership: |
|