Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00491175_m1
          See other G6PC2 GE Assays ›
          Gene Symbol:
          G6pc2
          Gene Name:
          glucose-6-phosphatase, catalytic, 2
          Gene Aliases:
          G6pc-rs, IGRP
          Chromosome Location:
          Chr.2: 69211073 - 69227993 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00491175_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: G6pc2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001289856.1 NP_001276785.1 2 - 3 189 58
          NM_001289857.1 NP_001276786.1 2 - 3 409 58
          NM_021331.4 NP_067306.1 2 - 3 409 58
          GenBank mRNA AK148450.1 - 2 - 3 410 58
          AK148465.1 - 2 - 3 361 58
          BC111905.1 - 2 - 3 384 58
          BQ557220.1 - 2 - 3 189 58
          Z47787.1 - 2 - 3 387 58

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200001003.mm.1

          Additional Information:

          This assay is in the 5'UTR region of the BC111905.1,NM_001289856.1 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Mus musculus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene G6pc2

          Panther Classification:

          Molecular Function -

          phosphatase

          Gene Ontology Categories:

          Function(s) Process(es)

          gluconeogenesis
          glucose homeostasis
          regulation of insulin secretion
          glucose 6-phosphate metabolic process
          catalytic activity
          glucose-6-phosphatase activity
          hydrolase activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0200007853.mm.1 NM_001289856.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200007853.mm.1 NM_001289857.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200007853.mm.1 NM_021331.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0200007853.mm.1 TC0200001003.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200007848.mm.1 NM_001289857.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200007848.mm.1 NM_021331.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0200007848.mm.1 TC0200001003.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200007846.mm.1 NM_001289856.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200007846.mm.1 TC0200001003.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200007851.mm.1 NM_001289857.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200007851.mm.1 NM_021331.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0200007851.mm.1 TC0200001003.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200007852.mm.1 NM_001289856.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200007852.mm.1 NM_001289857.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200007852.mm.1 NM_021331.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0200007852.mm.1 TC0200001003.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200007849.mm.1 NM_001289857.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200007849.mm.1 NM_021331.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0200007849.mm.1 TC0200001003.mm.1 MTA TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-8ltcg:80/100.66.77.141:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline