Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00491213_s1
          See other CHML GE Assays ›
          Gene Symbol:
          Chml
          Gene Name:
          choroideremia-like
          Gene Aliases:
          AI450814, E030003F13Rik, REP-2, Rep2
          Chromosome Location:
          Chr.1: 175682237 - 175688353 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00491213_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Chml
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_021350.2 NP_067325.2 1 1752 59
          GenBank mRNA BC118932.2 - 1 1752 40090670 59

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0100003612.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Chml

          Panther Classification:

          Molecular Function -

          G-protein modulator protein-binding activity modulator

          Gene Ontology Categories:

          Function(s) Process(es)

          intracellular protein transport
          protein geranylgeranylation
          GTPase activator activity
          Rab GTPase binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20849639 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20849638 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20849636 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1989782 BC118932.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1989782 NM_021350.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P113672 NM_021350.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P113672 AK086827.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P113672 NM_021350.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P113672 AK086827.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P126905 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P113672 NM_021350.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P113672 AK086827.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P113672 NM_021350.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P113672 AK086827.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1989782 BC118932.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1989782 NM_021350.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P382684 BC118932.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P382684 NM_021350.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20849624 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20849620 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20849633 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20849636 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20849637 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20849630 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20849626 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20849629 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20849622 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20849619 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20849639 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20849631 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20849634 - -
          Whole Mouse Genome G4122A A_51_P382684 BC118932.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P382684 NM_021350.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P330822 NM_021350.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P117625 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00593015 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00593033 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20849639 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20849636 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20849638 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20849637 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20849565 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20849666 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20849639 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20849661 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20849576 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20849636 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20849590 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20849566 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20849600 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20849563 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20849626 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20849662 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20849620 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20849587 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20849671 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20849560 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20849569 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20849633 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20849596 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20849648 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20849616 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20849557 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20849574 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20849602 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20849643 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20849599 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20849634 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20849614 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20849584 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20849655 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20849570 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20849622 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20849658 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20849609 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20849585 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20849631 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20849646 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20849567 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20849641 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20849603 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20849663 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20849572 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20849642 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20849610 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20849561 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20849650 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20849592 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20849583 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20849673 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20849558 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20849594 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20849579 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20849653 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20849612 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20849618 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20849629 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20849606 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20849624 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20849645 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20849664 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20849657 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20849568 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20849597 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20849660 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20849630 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20849669 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20849630 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20849583 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20849575 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20849606 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20849612 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20849568 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20849576 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20849624 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00593013 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20849634 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20849557 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00593032 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00688134 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20849614 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20849600 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20849563 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20849561 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20849602 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20849592 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00688120 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20849599 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20849603 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20849618 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20849573 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20849584 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20849558 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20849622 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20849580 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20849639 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20849633 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20849636 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20849631 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20849570 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20849594 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20849610 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20849597 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00593048 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20849588 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00106630 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20849569 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20849616 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20849611 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00011625 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20849560 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20849566 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20849572 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20849565 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20849574 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20849567 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20849596 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20849609 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20849629 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20849585 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10360462 - -
          Affymetrix Mouse Gene 1.0 ST Array 10360461 - -
          Affymetrix HT MG-430 PM Array Plate 1422322_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1435926_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10360460 - -
          Mouse Genome 430 2.0 Array 1422322_at - -
          Mouse Genome 430 2.0 Array 1422322_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 12663_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 12663_at - -
          Affymetrix Mouse Clariom S Array PSR0100029689.mm.1 NM_021350.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100029689.mm.1 NM_021350.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0100029689.mm.1 TC0100003612.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100029690.mm.1 NM_021350.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100029690.mm.1 NM_021350.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0100029690.mm.1 TC0100003612.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100029691.mm.1 NM_021350.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100029691.mm.1 NM_021350.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0100029691.mm.1 TC0100003612.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100029688.mm.1 NM_021350.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100029688.mm.1 NM_021350.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0100029688.mm.1 TC0100003612.mm.1 MTA TC
          Mouse Expression Set 430 1422322_PM_at - -
          Mouse Expression Set 430 1422322_at - -
          Mouse Genome 430A 2.0 Array 1422322_at - -
          Mouse Genome 430A 2.0 Array 1422322_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1422322_at - -
          Affymetrix HT Mouse Genome 430A Array 1422322_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1435926_at - -
          Affymetrix HT Mouse Genome 430B Array 1435926_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 339418 - -
          ABI Mouse Genome Survey Microarray 386372 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 6650156 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006650156 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2776066 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2776066 - -
          mouse-6 v1.1 (Illumina) scl15889.1.1_221-S - -
          Illumina Sentrix mouse-6.v1 bead array 6650156 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl15889.1.1_221-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6650156 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2776066 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-6wvps:80/100.66.75.98:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0