Search
Search

Mmp19| Gene Symbol: Mmp19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164197.1 | NP_001157669.1 | 2 - 3 | 142 | 81 | |
| NM_021412.2 | NP_067387.1 | 2 - 3 | 277 | 81 | ||
| GenBank mRNA | AF153199.1 | - | 2 - 3 | 229 | 81 | |
| AF155221.1 | - | 2 - 3 | 257 | 81 | ||
| AF162446.1 | - | 2 - 3 | 257 | 81 | ||
| AK079924.1 | - | 2 - 3 | 143 | 81 | ||
| AK080125.1 | - | 2 - 3 | 137 | 81 | ||
| AK136517.1 | - | 2 - 3 | 131 | 81 | ||
| AK154863.1 | - | 2 - 3 | 270 | 81 | ||
| BC112966.2 | - | 2 - 3 | 249 | 40039023 | 81 | |
Set Membership: |
|