Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 我們的服務對象
    • 生物技術
    • 生物製藥
    • CDMO
    • 實驗室診斷
    • 工業與應用科學
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00491465_s1
          See other SLC18A3 GE Assays ›
          Gene Symbol:
          Slc18a3
          Gene Name:
          solute carrier family 18 (vesicular monoamine), member 3
          Gene Aliases:
          VAChT, VAT
          Chromosome Location:
          Chr.14: 32462437 - 32464850 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00491465_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Slc18a3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_021712.2 NP_068358.2 1 1968 53
          GenBank mRNA AK158592.1 - 1 1970 53
          BC120498.1 - 1 1585 40129421 53
          BC137705.1 - 1 1584 8860724 53

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Transporter 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus Mouse Alzheimer's Disease 1700 Inventoried Mouse_Alzheimer Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Slc18a3

          Panther Classification:

          Molecular Function -

          secondary carrier transporter

          Gene Ontology Categories:

          Function(s) Process(es)

          cation transport
          neurotransmitter transport
          organic cation transport
          monoamine transport
          transmembrane transport
          ammonium transmembrane transport
          cation transmembrane transport
          acetate ester transport
          acetylcholine transmembrane transporter activity
          monoamine transmembrane transporter activity
          acetylcholine binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29517799 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29517814 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29517796 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29517817 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29517815 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29517801 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29517807 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29517794 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29517792 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29517802 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29517798 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29517809 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29517808 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29517813 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29517811 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29517806 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29517795 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29517812 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2179572 NM_021712.2 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P137603 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2179572 NM_021712.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P303675 BC137705.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P303675 NM_021712.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P303675 AK158592.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P303675 BC120498.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29517812 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29517817 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29517809 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29517811 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29517813 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29517806 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29517802 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29517808 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29517798 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29517820 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29517804 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29517810 - -
          Whole Mouse Genome G4122A A_51_P303675 BC137705.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P303675 NM_021712.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P303675 AK158592.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P303675 BC120498.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P128999 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11706673 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11332016 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11331999 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11332008 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11706671 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29517815 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29517806 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29517813 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29517808 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29517817 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29517794 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29517811 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29517801 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29517796 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29517814 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29517795 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29517807 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29517798 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29517812 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29517799 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29517792 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29517809 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29517802 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29517862 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29517837 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29517798 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29517806 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29517851 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29517875 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29517870 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29517843 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29517847 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29517793 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29517834 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29517872 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29517869 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29517850 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29517836 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29517828 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29517876 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29517878 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29517795 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29517845 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29517820 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29517873 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29517883 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29517827 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29517788 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29517790 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29517880 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29517829 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29517811 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29517797 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29517840 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29517864 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29517832 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29517817 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29517867 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29517823 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29517825 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29517810 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29517822 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29517804 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29517813 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29517855 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29517844 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29517808 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29517833 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29517830 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29517857 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29517791 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29517865 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29517860 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29517802 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29517809 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29517874 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29517798 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29517795 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29517791 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29517788 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29517793 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29517802 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29517813 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29517804 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29517806 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29517815 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29517797 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29517809 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29517820 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29517790 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29517817 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29517811 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10418837 - -
          Affymetrix Mouse Gene 1.0 ST Array 10418836 - -
          Affymetrix HT MG-430 PM Array Plate 1422203_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10418835 - -
          Mouse Genome 430 2.0 Array 1422203_PM_at - -
          Mouse Genome 430 2.0 Array 1422203_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 20508_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 20508_at - -
          Affymetrix Mouse Clariom S Array PSR1400013225.mm.1 NM_021712.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400013225.mm.1 NM_021712.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1400013225.mm.1 TC1400001704.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400013224.mm.1 NM_021712.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400013224.mm.1 NM_021712.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1400013224.mm.1 TC1400001704.mm.1 MTA TC
          Mouse Expression Set 430 1422203_at - -
          Mouse Expression Set 430 1422203_PM_at - -
          Murine Genome U74v2 Set 101354_at - -
          Murine Genome U74v2 Set 101354_PM_at - -
          Mouse Genome 430A 2.0 Array 1422203_PM_at - -
          Mouse Genome 430A 2.0 Array 1422203_at - -
          Affymetrix HT Mouse Genome 430A Array 1422203_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1422203_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 663017 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 3060035 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003060035 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1247491 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1247491 - -
          mouse-6 v1.1 (Illumina) scl45744.1.427_89-S - -
          Illumina Sentrix mouse-6.v1 bead array 3060035 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl45744.1.427_89-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3060035 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1247491 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1247491 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl45744.1.427_89-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-4lm45:80/100.66.75.98:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0