Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00491655_m1
          See other MYL7 GE Assays ›
          Gene Symbol:
          Myl7
          Gene Name:
          myosin, light polypeptide 7, regulatory
          Gene Aliases:
          MLC-2alpha, MLC2a, MYL2A, Mylc2a, RLC-A
          Chromosome Location:
          Chr.11: 5896637 - 5898782 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00491655_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Myl7
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_022879.2 NP_075017.2 4 - 5 317 76
          GenBank mRNA AJ348866.1 - 3 - 4 227 76
          AK146716.1 - 4 - 5 318 76
          BC061100.1 - 4 - 5 313 30300846 76
          S70785.1 - 3 - 4 262 76

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1100002156.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Myl7

          Panther Classification:

          Molecular Function -

          actin or actin-binding cytoskeletal protein

          Gene Ontology Categories:

          Process(es)

          calcium ion binding
          protein binding
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_65_P10195 S70785.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_65_P10195 NM_022879.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_65_P10195 AJ348866.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_65_P10195 AK146716.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_65_P10195 BC061100.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 853783 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P104189 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_65_P10195 S70785.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_65_P10195 NM_022879.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_65_P10195 AJ348866.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_65_P10195 AK146716.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_65_P10195 BC061100.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P246345 NM_022879.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P246345 AJ348866.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27565536 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27565547 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27565551 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27565557 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27565539 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27565528 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27565553 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27565544 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27565545 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27565533 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27565550 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27565524 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27565531 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27565538 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27565521 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27565546 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27565554 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27565525 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27565556 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27565526 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27565549 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27565534 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27565519 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27565535 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27565552 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27565529 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27565542 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P104189 - -
          Whole Mouse Genome G4122A A_51_P246345 NM_022879.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P246345 AJ348866.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P116792 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P101504 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P104189 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09103873 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09103879 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09103876 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09103874 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09103903 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09103902 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09103897 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09103887 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27565539 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27565500 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27565557 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27565524 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27565507 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27565467 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27565549 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27565479 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27565474 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27565493 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27565529 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27565463 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27565542 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27565488 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27565468 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27565498 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27565485 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27565510 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27565553 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27565477 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27565526 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27565556 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27565472 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27565487 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27565536 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27565519 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27565495 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27565521 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27565552 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27565547 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27565544 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27565504 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27565518 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27565550 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27565478 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27565511 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27565531 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27565525 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27565476 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27565515 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27565538 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27565506 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27565534 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27565546 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27565481 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27565551 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27565499 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27565517 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27565538 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09239671 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27565519 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09103904 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09115704 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27565511 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27565499 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27565525 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27565536 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27565502 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09103876 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27565544 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09103864 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27565556 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27565531 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27565503 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27565541 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27565534 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27565515 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27565493 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27565510 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09103903 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27565524 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09485801 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27565518 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27565539 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27565546 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09485802 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27565557 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27565504 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27565529 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27565535 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27565551 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27565513 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27565492 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09103896 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09103897 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27565553 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27565517 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09115702 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09078104 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09369616 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27565552 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27565498 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10384048 - -
          Affymetrix Mouse Gene 1.0 ST Array 10384049 - -
          Affymetrix Mouse Gene 1.0 ST Array 10384046 - -
          Affymetrix Mouse Gene 1.0 ST Array 10384050 - -
          Affymetrix Mouse Gene 1.0 ST Array 10384045 - -
          Affymetrix Mouse Gene 1.0 ST Array 10384047 - -
          Affymetrix HT MG-430 PM Array Plate 1449071_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10384044 - -
          Mouse Genome 430 2.0 Array 1449071_at - -
          Mouse Genome 430 2.0 Array 1449071_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 17898_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 17898_at - -
          Affymetrix Mouse Clariom S Array PSR1100020963.mm.1 NM_022879.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100020963.mm.1 NM_022879.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100020963.mm.1 TC1100002156.mm.1 MTA TC
          Mouse Expression Set 430 1449071_PM_at - -
          Mouse Expression Set 430 1449071_at - -
          Murine Genome U74v2 Set 100403_at - -
          Murine Genome U74v2 Set 100403_PM_at - -
          Murine Genome U74v2 Set 170292_at - -
          Murine Genome U74v2 Set 170292_PM_at - -
          Mouse Genome 430A 2.0 Array 1449071_at - -
          Mouse Genome 430A 2.0 Array 1449071_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1449071_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1449071_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 519196 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 2480541 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002480541 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2768253 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2768253 - -
          mouse-6 v1.1 (Illumina) scl017898.5_1-S - -
          Illumina Sentrix mouse-6.v1 bead array 2480541 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl017898.5_1-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2480541 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2768252 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2768252 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline