Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00491708_m1
          See other RCE1 GE Assays ›
          Gene Symbol:
          Rce1
          Gene Name:
          RCE1 homolog, prenyl protein peptidase (S. cerevisiae)
          Gene Aliases:
          D19Ertd283e, D19Ertd98e
          Chromosome Location:
          Chr.19: 4622551 - 4625617 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00491708_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rce1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_023131.1 NP_075620.1 4 - 5 453 74
          GenBank mRNA AJ251645.1 - 4 - 5 453 74
          AK012946.1 - 4 - 5 409 74
          BC004060.1 - 3 - 4 302 3590170 74

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1900000926.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Protease Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rce1

          Panther Classification:

          Molecular Function -

          metalloprotease

          Gene Ontology Categories:

          Function(s) Process(es)

          CAAX-box protein processing
          cysteine-type endopeptidase activity
          hydrolase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31924141 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31924139 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31924146 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31924143 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31924145 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31924140 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31924144 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31924147 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P130469 AK012946.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P130469 BC004060.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P130469 BC048686.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P130469 NM_023131.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P130469 AJ251645.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 641702 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 839470 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P104104 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P130469 AK012946.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P130469 BC004060.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P130469 BC048686.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P130469 NM_023131.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P130469 AJ251645.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P108778 NM_023131.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P108779 AK012946.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P108779 BC004060.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P108779 BC048686.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P108779 NM_023131.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31924147 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31924162 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31924141 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31924154 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31924160 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31924156 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31924125 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31924167 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31924172 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31924132 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31924158 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31924148 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31924139 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31924121 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31924144 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31924170 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31924138 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31924135 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31924142 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31924120 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31924128 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31924122 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31924131 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31924164 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31924151 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31924152 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31924149 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31924130 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31924119 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31924163 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31924174 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31924137 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31924123 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31924169 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31924129 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P104104 - -
          Whole Mouse Genome G4122A A_51_P108779 AK012946.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P108779 BC004060.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P108779 BC048686.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P108779 NM_023131.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P108778 NM_023131.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P117007 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P130469 AK012946.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P130469 BC004060.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P130469 BC048686.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P130469 NM_023131.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P130469 AJ251645.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P104104 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14409701 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14409691 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14409707 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14409670 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14409736 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14409722 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31924141 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31924147 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31924144 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31924146 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31924145 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31924139 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31924140 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31924143 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31924120 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31924121 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31924044 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31924034 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31924072 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31924170 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31924138 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31924165 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31924062 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31924113 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31924148 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31924115 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31924116 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31924091 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31924188 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31924058 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31924046 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31924078 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31924169 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31924050 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31924080 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31924164 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31924033 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31924066 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31924075 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31924043 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31924128 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31924152 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31924141 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31924061 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31924070 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31924079 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31924093 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31924176 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31924167 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31924040 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31924055 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31924076 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31924063 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31924088 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31924163 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31924130 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31924068 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31924119 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31924183 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31924144 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31924036 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31924132 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31924174 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31924056 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31924135 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31924125 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31924172 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31924129 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31924168 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31924149 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31924073 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31924086 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31924084 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31924131 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31924162 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31924137 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31924048 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31924064 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31924081 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31924057 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31924052 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31924042 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31924154 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31924122 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31924074 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31924113 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14211534 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31924149 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31924121 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31924178 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31924152 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31924128 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31924154 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31924135 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31924179 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31924160 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31924174 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14409745 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31924161 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31924116 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14409684 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31924172 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31924137 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31924158 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14214613 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31924123 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14214614 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31924144 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31924131 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31924148 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31924163 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14391076 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14268829 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31924165 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31924119 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31924115 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31924141 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14430272 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31924159 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31924132 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31924156 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31924122 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31924138 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31924125 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14211533 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14391079 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10464792 - -
          Affymetrix Mouse Gene 1.0 ST Array 10464787 - -
          Affymetrix Mouse Gene 1.0 ST Array 10464785 - -
          Affymetrix Mouse Gene 1.0 ST Array 10464790 - -
          Affymetrix Mouse Gene 1.0 ST Array 10464784 - -
          Affymetrix Mouse Gene 1.0 ST Array 10464786 - -
          Affymetrix Mouse Gene 1.0 ST Array 10464791 - -
          Affymetrix Mouse Gene 1.0 ST Array 10464788 - -
          Affymetrix Mouse Gene 1.0 ST Array 10464789 - -
          Affymetrix HT MG-430 PM Array Plate 1418779_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10464783 - -
          Mouse Genome 430 2.0 Array 1418779_at - -
          Mouse Genome 430 2.0 Array 1418779_PM_at - -
          Mouse Genome 430 2.0 Array 1429917_at - -
          Mouse Genome 430 2.0 Array 1429917_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 19671_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 19671_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1900008148.mm.1 NM_023131.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900008148.mm.1 NM_023131.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900008148.mm.1 TC1900000926.mm.1 MTA TC
          Mouse Expression Set 430 1418779_at - -
          Mouse Expression Set 430 1418779_PM_at - -
          Murine Genome U74v2 Set 96838_PM_at - -
          Murine Genome U74v2 Set 96838_at - -
          Mouse Genome 430A 2.0 Array 1418779_at - -
          Mouse Genome 430A 2.0 Array 1418779_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1418779_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1418779_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 325338 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 1980372 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001980372 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1235234 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1235234 - -
          mouse-6 v1.1 (Illumina) scl53464.6.1_108-S - -
          Illumina Sentrix mouse-6.v1 bead array 1980372 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl53464.6.1_108-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1980372 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1235234 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1235234 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl53464.6.1_108-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline