Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00492368_m1
          See other GNL1 GE Assays ›
          Gene Symbol:
          Gnl1
          Gene Name:
          guanine nucleotide binding protein-like 1
          Gene Aliases:
          Gna-rs1, Gnal1
          Chromosome Location:
          Chr.17: 35979955 - 35989462 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00492368_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gnl1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_008136.2 NP_032162.2 8 - 9 1314 71
          GenBank mRNA AK078187.1 - 8 - 9 1277 71
          BC010298.1 - 8 - 9 1269 3155411 71
          BC013243.1 - 8 - 9 1338 71
          BC094391.1 - 6 - 7 751 71
          BC110397.1 - 4 - 5 579 71
          BC129969.1 - 6 - 7 846 71
          BC129970.1 - 6 - 7 846 71
          X65026.1 - 7 - 8 956 71

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1700000715.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Gnl1

          Gene Ontology Categories:

          Function(s) Process(es)

          cellular response to DNA damage stimulus
          ribosome biogenesis
          nucleotide binding
          GTPase activity
          GTP binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31162035 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31162029 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31162032 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31162034 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31162036 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31162030 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31162031 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31162033 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31162038 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2177173 BC094391.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2177173 NM_008136.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2177173 AK078187.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2177173 AV338067.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 301778 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 581873 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 499397 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 874516 AK089546.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 874516 AK155267.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 874516 AK164544.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 874516 BC026718.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 874516 BC034249.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 874516 NM_198012.2 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 258379 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 766226 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 14807 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P171701 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2177173 BC094391.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2177173 NM_008136.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2177173 AK078187.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2177173 AV338067.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31162066 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31162069 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31162082 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31162057 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31162071 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31162062 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31162051 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31162059 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31162070 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31162061 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31162058 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31162080 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31162076 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31162053 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31162074 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31162073 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31162083 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31162047 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31162049 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31162068 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31162065 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31162055 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31162077 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31162064 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31162050 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P171701 - -
          Whole Mouse Genome G4122A A_52_P491766 X65026.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P491766 BC110397.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P491766 BC129969.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P491766 BC129970.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P491766 BC010298.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P102809 X65026.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P118660 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P171701 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13092236 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13092249 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13092246 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31162031 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31162032 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31162034 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31162036 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31162029 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31162030 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31162035 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31162038 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31162033 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31162139 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31162106 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31162012 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31162034 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31162147 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31162123 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31162146 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31162110 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31162023 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31162128 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31162022 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31162130 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31162127 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31162137 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31162140 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31162142 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31162021 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31162029 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31162036 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31162126 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31162111 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31162145 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31162113 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31162038 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31162025 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31162011 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31162143 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31162116 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31162031 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31162121 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31162108 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31162016 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31162018 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31162120 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31162114 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31162118 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31162119 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13258489 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31162134 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31162123 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13108030 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31162110 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13240784 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31162132 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31162022 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31162126 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31162143 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31162015 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13092238 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31162021 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31162011 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31162127 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13240785 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31162031 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31162142 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31162025 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31162145 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31162109 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13531355 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31162147 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31162018 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31162036 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31162118 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31162023 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13547675 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13203448 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13092804 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31162016 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31162130 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13238996 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13355892 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13092244 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31162020 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31162106 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31162140 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31162034 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31162124 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31162139 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13329427 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31162116 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31162108 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31162146 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31162128 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10444990 - -
          Affymetrix Mouse Gene 1.0 ST Array 10444988 - -
          Affymetrix Mouse Gene 1.0 ST Array 10444979 - -
          Affymetrix Mouse Gene 1.0 ST Array 10444981 - -
          Affymetrix Mouse Gene 1.0 ST Array 10444987 - -
          Affymetrix Mouse Gene 1.0 ST Array 10444986 - -
          Affymetrix Mouse Gene 1.0 ST Array 10444982 - -
          Affymetrix Mouse Gene 1.0 ST Array 10444985 - -
          Affymetrix Mouse Gene 1.0 ST Array 10444984 - -
          Affymetrix Mouse Gene 1.0 ST Array 10444989 - -
          Affymetrix Mouse Gene 1.0 ST Array 10444983 - -
          Affymetrix Mouse Gene 1.0 ST Array 10444980 - -
          Affymetrix HT MG-430 PM Array Plate 1418647_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1418646_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10444978 - -
          Mouse Genome 430 2.0 Array 1418646_PM_at - -
          Mouse Genome 430 2.0 Array 1418646_at - -
          Mouse Genome 430 2.0 Array 1418647_at - -
          Mouse Genome 430 2.0 Array 1418647_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 14670_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 14670_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1700007375.mm.1 NM_008136.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700007375.mm.1 NM_008136.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700007375.mm.1 TC1700000715.mm.1 MTA TC
          Mouse Expression Set 430 1418646_PM_at - -
          Mouse Expression Set 430 1418647_at - -
          Mouse Expression Set 430 1418647_PM_at - -
          Mouse Expression Set 430 1418646_at - -
          Murine Genome U74v2 Set 98403_at - -
          Murine Genome U74v2 Set 161304_PM_r_at - -
          Murine Genome U74v2 Set 98403_PM_at - -
          Murine Genome U74v2 Set 161304_r_at - -
          Mouse Genome 430A 2.0 Array 1418646_at - -
          Mouse Genome 430A 2.0 Array 1418647_PM_at - -
          Mouse Genome 430A 2.0 Array 1418646_PM_at - -
          Mouse Genome 430A 2.0 Array 1418647_at - -
          Affymetrix HT Mouse Genome 430A Array 1418646_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1418646_at - -
          Affymetrix HT Mouse Genome 430A Array 1418647_at - -
          Affymetrix HT Mouse Genome 430A Array 1418647_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 843747 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 104200020 - -
          Illumina mouse-6 v1.0 expression beadchip 104200020 - -
          Illumina mouse-6 v1.0 expression beadchip 6660609 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006660609 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1223648 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1223648 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1230243 - -
          mouse-6 v1.1 (Illumina) scl0014670.2_304-S - -
          mouse-6 v1.1 (Illumina) scl50754.6_255-S - -
          Illumina Sentrix mouse-6.v1 bead array 104200020 - -
          Illumina Sentrix mouse-6.v1 bead array 6660609 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0014670.2_304-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6660609 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 104200020 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2932824 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2932824 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1223648 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1230243 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl0014670.2_304-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline