Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00492601_m1
          See other IFI203 GE Assays ›
          Gene Symbol:
          Ifi203
          Gene Name:
          interferon activated gene 203
          Gene Aliases:
          -
          Chromosome Location:
          Chr.1: 173920400 - 173942847 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00492601_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ifi203
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AF022371.1 - 1157 132
          AK040214.1 - 6 - 7 1097 132
          AK137616.1 - 8 - 9 2482 132
          AK144663.1 - 7 - 8 1222 132
          AK164695.1 - 4 - 5 934 132
          AK169754.1 - 6 - 7 1091 132
          BB206743.2 - 1 - 2 306 132
          BC008167.1 - 1343 3257492 132
          L14559.1 - 3 - 4 733 132

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0100003891.mm.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Mus musculus Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          DNA-binding transcription factor

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2004507 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1966731 L14559.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1966731 BC008167.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1966731 BB206743.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1966731 AK164695.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1966731 AK169754.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1998843 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1988202 BF454130.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1988202 NM_008328.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1988202 NM_001045481.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P155880 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P178322 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P145095 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P173111 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1988202 BF454130.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1988202 NM_008328.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1988202 NM_001045481.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2004507 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1998843 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1966731 L14559.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1966731 BC008167.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1966731 BB206743.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1966731 AK164695.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1966731 AK169754.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20840123 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20840121 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04359788 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04359771 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P178322 - -
          Whole Mouse Genome G4122A A_52_P134762 BC057140.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P134762 AK172505.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P134762 AK041232.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P237856 BF454130.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P237856 AK144663.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P237856 AF022371.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P237856 NM_008328.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P237856 NM_001045481.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P263800 AK080088.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P137967 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P178322 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20839957 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20839970 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20839960 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20839946 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20839945 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20839990 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20839963 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20839941 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20839973 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20839968 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20839949 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20839974 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20839978 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20839992 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20839972 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20840123 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20839987 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20839944 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00917667 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20839974 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20840123 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20839975 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20839970 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20840074 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20840093 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20839973 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20840067 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20839972 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20840121 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10360396 - -
          Affymetrix Mouse Gene 1.0 ST Array 10360393 - -
          Affymetrix Mouse Gene 1.0 ST Array 10360397 - -
          Affymetrix Mouse Gene 1.0 ST Array 10360394 - -
          Affymetrix Mouse Gene 1.0 ST Array 10360392 - -
          Affymetrix Mouse Gene 1.0 ST Array 10360395 - -
          Affymetrix HT MG-430 PM Array Plate 1451567_PM_a_at - -
          Affymetrix HT MG-430 PM Array Plate 1448775_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1425008_PM_a_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10360391 - -
          Mouse Genome 430 2.0 Array 1425008_PM_a_at - -
          Mouse Genome 430 2.0 Array 1451567_PM_a_at - -
          Mouse Genome 430 2.0 Array 1451567_a_at - -
          Mouse Genome 430 2.0 Array 1452348_s_at - -
          Mouse Genome 430 2.0 Array 1425008_a_at - -
          Mouse Genome 430 2.0 Array 1442828_at - -
          Mouse Genome 430 2.0 Array 1448775_at - -
          Mouse Genome 430 2.0 Array 1442828_PM_at - -
          Mouse Genome 430 2.0 Array 1452348_PM_s_at - -
          Mouse Genome 430 2.0 Array 1448775_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 15950_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 15950_at - -
          Affymetrix Mouse Clariom S Array PSR0100029572.mm.1 TC0100003891.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100029569.mm.1 TC0100003891.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100029570.mm.1 TC0100003891.mm.1 MTA TC
          Mouse Expression Set 430 1425008_PM_a_at - -
          Mouse Expression Set 430 1448775_PM_at - -
          Mouse Expression Set 430 1451567_a_at - -
          Mouse Expression Set 430 1452348_s_at - -
          Mouse Expression Set 430 1448775_at - -
          Mouse Expression Set 430 1451567_PM_a_at - -
          Mouse Expression Set 430 1425008_a_at - -
          Mouse Expression Set 430 1452348_PM_s_at - -
          Mouse Expression Set 430 1442828_PM_at - -
          Mouse Expression Set 430 1442828_at - -
          Murine Genome U74v2 Set 93321_at - -
          Murine Genome U74v2 Set 93321_PM_at - -
          Murine Genome U74v2 Set 94224_s_at - -
          Murine Genome U74v2 Set 94224_PM_s_at - -
          Mouse Genome 430A 2.0 Array 1452348_s_at - -
          Mouse Genome 430A 2.0 Array 1425008_a_at - -
          Mouse Genome 430A 2.0 Array 1452348_PM_s_at - -
          Mouse Genome 430A 2.0 Array 1448775_PM_at - -
          Mouse Genome 430A 2.0 Array 1451567_a_at - -
          Mouse Genome 430A 2.0 Array 1451567_PM_a_at - -
          Mouse Genome 430A 2.0 Array 1448775_at - -
          Mouse Genome 430A 2.0 Array 1425008_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1426906_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1425008_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1452231_PM_x_at - -
          Affymetrix HT Mouse Genome 430A Array 1426906_at - -
          Affymetrix HT Mouse Genome 430A Array 1448775_at - -
          Affymetrix HT Mouse Genome 430A Array 1425008_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1451567_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1448775_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1451567_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1452231_x_at - -
          Affymetrix HT Mouse Genome 430B Array 1442828_at - -
          Affymetrix HT Mouse Genome 430B Array 1442828_PM_at - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 6420102 - -
          Illumina mouse-6 v1.0 expression beadchip 3610369 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006420102 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003610369 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1241443 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1241443 - -
          mouse-6 v1.1 (Illumina) scl0015950.1_6-S - -
          mouse-6 v1.1 (Illumina) scl000987.1_58-S - -
          Illumina Sentrix mouse-6.v1 bead array 6420102 - -
          Illumina Sentrix mouse-6.v1 bead array 3610369 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl000987.1_58-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0015950.1_6-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3610369 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6420102 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1241443 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1224389 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1224389 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1241443 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl0015950.1_6-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl000987.1_58-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline