Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00492707_m1
          See other ITGB1BP1 GE Assays ›
          Gene Symbol:
          Itgb1bp1
          Gene Name:
          integrin beta 1 binding protein 1
          Gene Aliases:
          AI449260, AU019480, Icap1
          Chromosome Location:
          Chr.12: 21267246 - 21286279 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00492707_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Itgb1bp1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_008403.4 NP_032429.1 4 - 5 478 77
          GenBank mRNA AJ001373.1 - 4 - 5 460 77
          AK002786.1 - 4 - 5 435 77
          AK004172.1 - 4 - 5 434 77
          AK007635.1 - 4 - 5 436 77
          AK075577.1 - 4 - 5 479 77
          AK089937.1 - 4 - 5 592 77
          AK147929.1 - 4 - 5 427 77
          BC021151.1 - 4 - 5 617 77
          BC028772.1 - 3 - 4 314 1230640 77

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1200001489.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Itgb1bp1

          Gene Ontology Categories:

          Function(s) Process(es)

          angiogenesis
          blood vessel endothelial cell proliferation involved in sprouting angiogenesis
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          negative regulation of protein kinase activity
          negative regulation of cell adhesion involved in substrate-bound cell migration
          cell adhesion
          cell-matrix adhesion
          Notch signaling pathway
          integrin-mediated signaling pathway
          positive regulation of cell proliferation
          negative regulation of cell proliferation
          positive regulation of endothelial cell migration
          negative regulation of fibroblast migration
          protein transport
          cell differentiation
          regulation of cell adhesion
          biomineral tissue development
          negative regulation of protein binding
          activation of protein kinase B activity
          integrin activation
          regulation of cell adhesion mediated by integrin
          tube formation
          cellular response to vascular endothelial growth factor stimulus
          regulation of GTPase activity
          receptor clustering
          cellular response to fibroblast growth factor stimulus
          positive regulation of Notch signaling pathway
          positive regulation of transcription from RNA polymerase II promoter
          regulation of blood vessel size
          myoblast migration
          positive regulation of stress fiber assembly
          positive regulation of cell division
          positive regulation of focal adhesion assembly
          negative regulation of focal adhesion assembly
          positive regulation of protein kinase B signaling
          negative regulation of ERK1 and ERK2 cascade
          negative regulation of cell migration involved in sprouting angiogenesis
          positive regulation of protein targeting to membrane
          negative regulation of protein targeting to membrane
          negative regulation of substrate adhesion-dependent cell spreading
          regulation of integrin-mediated signaling pathway
          GDP-dissociation inhibitor activity
          integrin binding
          protein binding
          protein C-terminus binding
          protein transporter activity
          kinase binding
          protein kinase binding
          protein complex binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28279800 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28279804 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28279802 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28279803 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28279801 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P106363 AK147929.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P106363 BC021151.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P106363 BC028772.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P106363 AK002786.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P106363 AK004172.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P439267 BC038615.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P439267 BC080847.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P439267 BC096022.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P439267 NM_001004364.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P439267 NM_001098168.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P439267 BC038615.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P439267 BC080847.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P439267 BC096022.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P439267 NM_001004364.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P439267 NM_001098168.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 751818 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 742905 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 178278 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 671961 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 506795 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 680396 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 89324 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 872433 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 664496 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 53187 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P439267 BC038615.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P439267 BC080847.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P439267 BC096022.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P439267 NM_001004364.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P439267 NM_001098168.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P439267 BC038615.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P439267 BC080847.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P439267 BC096022.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P439267 NM_001004364.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P439267 NM_001098168.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P106363 AK147929.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P106363 BC021151.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P106363 BC028772.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P106363 AK002786.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P106363 AK004172.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28279782 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28279785 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28279778 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28279774 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28279780 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28279789 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28279784 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28279776 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28279797 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28279791 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28279800 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28279803 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28279796 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28279790 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28279792 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28279799 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28279788 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07556665 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07556651 - -
          Whole Mouse Genome G4122A A_52_P439267 BC038615.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P439267 BC080847.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P439267 BC096022.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P439267 NM_001004364.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P439267 NM_001098168.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P439267 BC038615.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P439267 BC080847.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P439267 BC096022.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P439267 NM_001004364.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P439267 NM_001098168.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P456980 AJ001373.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P106363 AK147929.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P106363 BC021151.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P106363 BC028772.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P106363 AK002786.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P106363 AK004172.1 mouse_transcriptome
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28279803 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28279802 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28279801 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28279800 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28279804 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28279747 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28279800 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28279722 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28279780 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28279793 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28279773 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28279768 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28279770 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28279729 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28279724 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28279711 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28279733 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28279726 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28279792 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28279737 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28279762 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28279742 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28279790 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28279709 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28279766 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28279713 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28279721 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28279782 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28279797 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28279758 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28279718 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28279784 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28279743 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28279741 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28279744 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28279719 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28279760 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28279755 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28279735 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28279803 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28279796 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28279717 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28279732 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28279788 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28279751 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28279728 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28279799 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28279752 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28279776 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28279768 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28279801 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28279785 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28279789 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28279790 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28279792 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28279793 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28279796 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28279767 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28279777 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28279799 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28279782 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28279773 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28279784 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28279778 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28279770 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28279803 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28279780 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28279797 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28279771 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28279788 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10399595 - -
          Affymetrix Mouse Gene 1.0 ST Array 10399601 - -
          Affymetrix Mouse Gene 1.0 ST Array 10399597 - -
          Affymetrix Mouse Gene 1.0 ST Array 10399600 - -
          Affymetrix Mouse Gene 1.0 ST Array 10399599 - -
          Affymetrix Mouse Gene 1.0 ST Array 10399603 - -
          Affymetrix Mouse Gene 1.0 ST Array 10399594 - -
          Affymetrix Mouse Gene 1.0 ST Array 10399598 - -
          Affymetrix Mouse Gene 1.0 ST Array 10399604 - -
          Affymetrix Mouse Gene 1.0 ST Array 10399602 - -
          Affymetrix Mouse Gene 1.0 ST Array 10399596 - -
          Affymetrix HT MG-430 PM Array Plate 1422157_PM_a_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10394862 - -
          Affymetrix Mouse Gene 1.1 ST Array 10399593 - -
          Mouse Genome 430 2.0 Array 1422157_a_at - -
          Mouse Genome 430 2.0 Array 1422157_PM_a_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 16413_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 16413_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1200010425.mm.1 NM_008403.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200010425.mm.1 NM_008403.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1200010425.mm.1 TC1200001489.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200010424.mm.1 NM_008403.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200010424.mm.1 NM_008403.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1200010424.mm.1 TC1200001489.mm.1 MTA TC
          Mouse Expression Set 430 1422157_PM_a_at - -
          Mouse Expression Set 430 1422157_a_at - -
          Murine Genome U74v2 Set 100991_at - -
          Murine Genome U74v2 Set 100989_PM_at - -
          Murine Genome U74v2 Set 100989_at - -
          Murine Genome U74v2 Set 100990_g_at - -
          Murine Genome U74v2 Set 100990_PM_g_at - -
          Murine Genome U74v2 Set 100991_PM_at - -
          Murine Genome U74v2 Set 170272_PM_r_at - -
          Murine Genome U74v2 Set 170272_r_at - -
          Mouse Genome 430A 2.0 Array 1422157_a_at - -
          Mouse Genome 430A 2.0 Array 1422157_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1422157_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1422157_a_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 637885 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 3830121 - -
          Illumina mouse-6 v1.0 expression beadchip 3830121 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003830121 - -
          mouse-6 v1.1 (Illumina) scl016413.7_44-S - -
          Illumina Sentrix mouse-6.v1 bead array 3830121 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl016413.7_44-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3830121 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2924243 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2924243 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-6wvps:80/100.66.75.98:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0