Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00492796_m1
          See other KCND1 GE Assays ›
          Gene Symbol:
          Kcnd1
          Gene Name:
          potassium voltage-gated channel, Shal-related family, member 1
          Gene Aliases:
          1110037K09Rik, Kca2-1, Kv4.1, Shal, Shal1, mShal1
          Chromosome Location:
          Chr.X: 7823843 - 7836503 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00492796_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Kcnd1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK033805.1 - 1288 110
          BC139325.1 - 5 - 6 1803 8862347 110
          BC139327.1 - 5 - 6 1803 8862349 110
          M64226.1 - 5 - 6 1719 110

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0X00000072.mm.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          voltage-gated ion channel

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1965478 BC139325.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1965478 BC139327.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1965478 M64226.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1965478 NM_008423.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1965478 AK033805.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P126157 AK033805.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 542547 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 579060 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 254283 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 591688 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 223174 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 569861 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 318102 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 79009 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P113022 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P153334 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P126157 AK033805.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1965478 BC139325.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1965478 BC139327.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1965478 M64226.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1965478 NM_008423.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1965478 AK033805.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2143976 - -
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P236053 BC139325.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P236053 BC139327.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P236053 M64226.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P236053 NM_008423.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P236053 AK033805.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32240340 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32240343 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32240368 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32240350 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32240355 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32240345 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32240351 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32240346 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32240363 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32240352 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32240353 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32240357 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32240362 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32240356 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32240372 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32240411 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32240369 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32240359 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32240367 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32240364 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32240365 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32240341 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32240375 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32240348 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32240371 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32240374 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32240360 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09226886 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P113022 - -
          Whole Mouse Genome G4122A A_51_P236053 BC139325.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P236053 BC139327.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P236053 M64226.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P236053 NM_008423.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P236053 AK033805.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P126157 AK033805.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P100064 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P113022 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P15084752 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P15084749 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14838362 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P15084748 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14838374 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P15084747 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32240343 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32240466 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32240468 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32240380 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32240458 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32240355 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32240446 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32240408 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32240475 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32240413 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32240431 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32240394 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32240397 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32240439 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32240472 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32240345 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32240350 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32240351 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32240374 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32240476 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32240406 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32240360 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32240424 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32240377 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32240456 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32240460 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32240447 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32240368 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32240353 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32240461 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32240400 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32240418 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32240438 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32240451 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32240459 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32240462 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32240385 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32240440 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32240420 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32240470 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32240348 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32240389 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32240362 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32240402 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32240443 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32240375 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32240356 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32240371 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32240391 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32240388 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32240379 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32240395 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32240382 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32240405 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32240372 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32240464 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32240399 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32240364 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32240434 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32240359 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32240416 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32240426 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32240473 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32240445 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32240393 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32240450 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32240437 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32240448 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32240463 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32240414 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32240367 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32240352 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32240411 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32240452 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32240421 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32240454 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14907407 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32240389 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32240377 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32240343 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32240371 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32240345 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P15102578 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P15102579 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32240356 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32240391 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32240353 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P15084748 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14907406 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14838372 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32240362 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32240409 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32240365 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32240367 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32240346 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32240364 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32240341 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14849963 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32240411 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32240375 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32240380 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32240359 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32240350 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32240379 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32240408 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32240368 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32240394 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32240360 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32240388 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32240373 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32240382 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32240395 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14849962 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32240372 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32240348 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32240393 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10598450 - -
          Affymetrix Mouse Gene 1.0 ST Array 10598454 - -
          Affymetrix Mouse Gene 1.0 ST Array 10598449 - -
          Affymetrix Mouse Gene 1.0 ST Array 10598452 - -
          Affymetrix Mouse Gene 1.0 ST Array 10598453 - -
          Affymetrix Mouse Gene 1.0 ST Array 10598451 - -
          Affymetrix Mouse Gene 1.0 ST Array 10598455 - -
          Affymetrix HT MG-430 PM Array Plate 1455514_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1446556_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1421538_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10598448 - -
          Mouse Genome 430 2.0 Array 1446556_at - -
          Mouse Genome 430 2.0 Array 1455514_at - -
          Mouse Genome 430 2.0 Array 1455514_PM_at - -
          Mouse Genome 430 2.0 Array 1421538_at - -
          Mouse Genome 430 2.0 Array 1421538_PM_at - -
          Mouse Genome 430 2.0 Array 1446556_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 16506_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 16506_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0X00000519.mm.1 NM_008423.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00000519.mm.1 TC0X00000072.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00000520.mm.1 TC0X00000072.mm.1 MTA TC
          Mouse Expression Set 430 1421538_PM_at - -
          Mouse Expression Set 430 1421538_at - -
          Mouse Expression Set 430 1455514_PM_at - -
          Mouse Expression Set 430 1455514_at - -
          Mouse Expression Set 430 1446556_PM_at - -
          Mouse Expression Set 430 1446556_at - -
          Murine Genome U74v2 Set 101302_PM_at - -
          Murine Genome U74v2 Set 101302_at - -
          Murine Genome U74v2 Set 139291_at - -
          Murine Genome U74v2 Set 139291_PM_at - -
          Mouse Genome 430A 2.0 Array 1421538_at - -
          Mouse Genome 430A 2.0 Array 1421538_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1421538_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1421538_at - -
          Affymetrix HT Mouse Genome 430B Array 1455514_at - -
          Affymetrix HT Mouse Genome 430B Array 1455514_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1446556_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1446556_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 457668 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 6860278 - -
          Illumina mouse-6 v1.0 expression beadchip 100870286 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006860278 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1226720 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1241328 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1226720 - -
          mouse-6 v1.1 (Illumina) scl0016506.2_100-S - -
          mouse-6 v1.1 (Illumina) scl55070.6_596-S - -
          Illumina Sentrix mouse-6.v1 bead array 100870286 - -
          Illumina Sentrix mouse-6.v1 bead array 6860278 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0016506.2_100-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6860278 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 100870286 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2726857 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2726857 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1241328 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline