Search
Search
| Gene Symbol: Lag3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008479.2 | NP_032505.1 | 3 - 4 | 855 | 113 | |
| GenBank mRNA | BC120591.1 | - | 3 - 4 | 787 | 40129514 | 113 |
| X98113.1 | - | 3 - 4 | 857 | 113 | ||
Set Membership: |
|