| Array Name |
Probe IDs |
Targets |
Target Type |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P248044 |
CK634059.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P248044 |
BC026051.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P248044 |
NM_008483.3 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P248044 |
CK634059.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P248044 |
BC026051.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P248044 |
NM_008483.3 |
mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
555410 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
273746 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
284567 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
739272 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
693212 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
839921 |
AK004557.2 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
839921 |
AK014681.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
839921 |
AK018432.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
839921 |
AK033733.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
839921 |
AK045596.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
839921 |
AK080201.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
839921 |
AK164665.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
839921 |
BC019452.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
839921 |
NM_025809.4 |
refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
129397 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
124557 |
- |
- |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P248044 |
CK634059.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P248044 |
BC026051.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P248044 |
NM_008483.3 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P248044 |
CK634059.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P248044 |
BC026051.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P248044 |
NM_008483.3 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P248044 |
CK634059.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P248044 |
BC026051.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P248044 |
NM_008483.3 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P248044 |
CK634059.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P248044 |
BC026051.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P248044 |
NM_008483.3 |
mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P09561288 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P09561300 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P09561335 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P09561319 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P09561310 |
- |
- |
| Whole Mouse Genome G4122A |
A_51_P248044 |
CK634059.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P248044 |
BC026051.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P248044 |
NM_008483.3 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P248044 |
CK634059.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P248044 |
BC026051.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P248044 |
NM_008483.3 |
mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P139031 |
- |
- |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_67_P09561310 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26815082 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26815062 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26815063 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26815087 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26815101 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26815075 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26815105 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26815057 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26815111 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26815086 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26815052 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26815046 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26815108 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26815051 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26815049 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26815097 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26815085 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26815113 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26815070 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26815109 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26815099 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26815091 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26815065 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26815081 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26815055 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26815076 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26815095 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26815072 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26815061 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26815103 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26815102 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26815079 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26815078 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26815069 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26815047 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P32975177 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P32975128 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P32975102 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P32975062 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P32975167 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P32975077 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P32975161 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P32975104 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P32975092 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P32975171 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P32975124 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P32975084 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P32975170 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P32975075 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P32975061 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P32975157 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P32975097 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P32975116 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P32975154 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P32975111 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P32975140 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P32975144 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P32975131 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P32975109 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P32975127 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P32975059 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P32975090 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P32975175 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P32975100 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P32975138 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P32975120 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P32975173 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P32975155 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P32975114 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P32975066 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P32975130 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P32975070 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P32975106 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P32975112 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P32975150 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P32975055 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P32975086 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P32975072 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P32975148 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P32975110 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P32975159 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P32975083 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P32975060 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P32975056 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P32975107 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P32975095 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P32975141 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P32975135 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P32975146 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P32975172 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P32975133 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P32975079 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P32975152 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P32975122 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P32975053 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P32975168 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P32975068 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P32975065 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P32975118 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P32975074 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P32975057 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P32975147 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P32975151 |
- |
- |