Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00493277_g1
          See other TULP2 GE Assays ›
          Gene Symbol:
          Tulp2
          Gene Name:
          tubby-like protein 2
          Gene Aliases:
          Pdet
          Chromosome Location:
          Chr.7: 45482026 - 45523712 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00493277_g1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tulp2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AF105712.1 - 12 - 13 1561 75
          AK054304.1 - 9 - 10 1539 75
          AK077200.1 - 12 - 13 1570 75
          AK077211.1 - 9 - 10 1367 75
          AK161165.1 - 9 - 10 1518 75
          BC048740.1 - 5 - 6 700 75
          BC089545.1 - 9 - 10 1366 6770730 75
          CD422160.1 - 2 - 3 267 75
          X69827.1 - 6 - 7 823 75

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700000849.mm.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2054132 X69827.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2054132 BC048740.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2054132 AK161165.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2054132 BC089545.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2054132 AK054304.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 576252 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 777670 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 176485 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 683333 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 401922 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 86763 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2054132 X69827.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2054132 BC048740.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2054132 AK161165.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2054132 BC089545.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2054132 AK054304.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25093599 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25093596 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25093594 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25093598 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25093603 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25093601 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25093602 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25093606 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25093604 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06183209 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06183193 - -
          Whole Mouse Genome G4122A A_51_P389017 X69827.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P133944 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P118509 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06402749 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06402752 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06402753 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06402764 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06402748 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06402757 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093560 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093610 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093659 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093547 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093571 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093627 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093563 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093650 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093562 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093525 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093661 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093549 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093505 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093638 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093594 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093633 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093585 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093568 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093535 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093606 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093648 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093519 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093598 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093551 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093623 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093604 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093501 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093615 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093545 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093646 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093530 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093608 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093540 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093558 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093566 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093555 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093660 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093564 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093626 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093637 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093614 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093665 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093596 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093587 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093656 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093521 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093574 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093593 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093612 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093653 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093583 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093588 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093516 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093529 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093628 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093514 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093536 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093533 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093534 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093591 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093526 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093498 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093543 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093631 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093553 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093576 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093565 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093580 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093602 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093635 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093520 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093506 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093523 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093503 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093548 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093590 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093554 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093664 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093640 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093557 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25093580 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06490017 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25093560 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25093548 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25093584 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05886715 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25093588 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25093566 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25093553 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25093578 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25093565 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25093558 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06119986 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25093544 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25093564 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25093596 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25093540 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25093568 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25093598 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06431900 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25093606 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25093571 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25093563 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25093555 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25093585 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06402755 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25093574 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06402748 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25093542 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25093557 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06490020 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25093594 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25093549 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05886717 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06402758 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06490023 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25093583 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06257216 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25093554 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25093590 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25093562 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06237345 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25093602 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25093543 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10552974 - -
          Affymetrix Mouse Gene 1.0 ST Array 10552967 - -
          Affymetrix Mouse Gene 1.0 ST Array 10552976 - -
          Affymetrix Mouse Gene 1.0 ST Array 10552981 - -
          Affymetrix Mouse Gene 1.0 ST Array 10552978 - -
          Affymetrix Mouse Gene 1.0 ST Array 10552969 - -
          Affymetrix Mouse Gene 1.0 ST Array 10552968 - -
          Affymetrix Mouse Gene 1.0 ST Array 10552975 - -
          Affymetrix Mouse Gene 1.0 ST Array 10552971 - -
          Affymetrix Mouse Gene 1.0 ST Array 10552977 - -
          Affymetrix Mouse Gene 1.0 ST Array 10552972 - -
          Affymetrix Mouse Gene 1.0 ST Array 10552980 - -
          Affymetrix Mouse Gene 1.0 ST Array 10552979 - -
          Affymetrix Mouse Gene 1.0 ST Array 10552973 - -
          Affymetrix Mouse Gene 1.0 ST Array 10552970 - -
          Affymetrix HT MG-430 PM Array Plate 1417276_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10552966 - -
          Mouse Genome 430 2.0 Array 1417276_PM_at - -
          Mouse Genome 430 2.0 Array 1417276_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 56734_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 56734_at - -
          Affymetrix Mouse Clariom S Array PSR0700007350.mm.1 TC0700000849.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700007350.mm.1 NM_001045555.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700007350.mm.1 NM_008807.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0700007351.mm.1 TC0700000849.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700007351.mm.1 NM_008807.3 Refseq
          Mouse Expression Set 430 1417276_PM_at - -
          Mouse Expression Set 430 1417276_at - -
          Murine Genome U74v2 Set 160863_PM_at - -
          Murine Genome U74v2 Set 160863_at - -
          Mouse Genome 430A 2.0 Array 1417276_at - -
          Mouse Genome 430A 2.0 Array 1417276_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1417276_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1417276_at - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 102370270 - -
          Illumina mouse-6 v1.0 expression beadchip 6420039 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006420039 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1256080 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1254263 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1256080 - -
          mouse-6 v1.1 (Illumina) scl000104.1_52-S - -
          mouse-6 v1.1 (Illumina) scl32679.14.1_2-S - -
          Illumina Sentrix mouse-6.v1 bead array 6420039 - -
          Illumina Sentrix mouse-6.v1 bead array 102370270 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl32679.14.1_2-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 102370270 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6420039 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1256080 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1256080 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1254263 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline