| Array Name |
Probe IDs |
Targets |
Target Type |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P275454 |
J03776.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P275454 |
BC005447.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P275454 |
DV644722.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P275454 |
AK137592.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P275454 |
AF220014.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P275454 |
J03776.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P275454 |
BC005447.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P275454 |
DV644722.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P275454 |
AK137592.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P275454 |
AF220014.1 |
mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
380979 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
344247 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
342963 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
40426 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
795163 |
AK036773.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
737433 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
451704 |
AK009767.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
451704 |
AK010267.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
451704 |
NM_007475.4 |
refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
318299 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
360220 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
535566 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
672805 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
156008 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
265449 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
275087 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
503030 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
859013 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
207076 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
541434 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
522220 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
162131 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
18581 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
146007 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
113798 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
154365 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
624 |
- |
- |
| Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P110316 |
- |
- |
| Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P140926 |
- |
- |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P275454 |
J03776.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P275454 |
BC005447.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P275454 |
DV644722.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P275454 |
AK137592.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P275454 |
AF220014.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P275454 |
J03776.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P275454 |
BC005447.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P275454 |
DV644722.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P275454 |
AK137592.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P275454 |
AF220014.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P275454 |
J03776.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P275454 |
BC005447.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P275454 |
DV644722.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P275454 |
AK137592.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P275454 |
AF220014.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P275454 |
J03776.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P275454 |
BC005447.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P275454 |
DV644722.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P275454 |
AK137592.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P275454 |
AF220014.1 |
mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25388246 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25388243 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25388252 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25388232 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25388234 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P06309033 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P06309075 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P06309093 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P06309066 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P01595182 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P110316 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P140926 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P01595169 |
- |
- |
| Whole Mouse Genome G4122A |
A_51_P275454 |
J03776.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P275454 |
BC005447.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P275454 |
DV644722.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P275454 |
AK137592.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P275454 |
AF220014.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P275454 |
J03776.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P275454 |
BC005447.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P275454 |
DV644722.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P275454 |
AK137592.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P275454 |
AF220014.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P442567 |
J03776.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P442567 |
BC005447.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P442567 |
AK137506.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P442567 |
AF220014.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P442567 |
NM_009099.2 |
mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P109621 |
- |
- |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P136542 |
- |
- |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P110517 |
- |
- |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P110316 |
- |
- |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P140926 |
- |
- |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_67_P06309066 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P06000421 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P06405728 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P06338532 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P06338535 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25388102 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25388193 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25388112 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25388088 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25388143 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25388063 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25388155 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25388072 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25388147 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25388184 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25388142 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25388176 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25388246 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25388086 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25388182 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25388119 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25388110 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25388153 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25388065 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25388125 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25388179 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25388227 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25388211 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25388105 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25388268 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25388104 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25388163 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25388157 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25388192 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25388174 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25388221 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25388185 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25388189 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25388084 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25388243 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25388099 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25388120 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25388201 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25388146 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25388195 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25388141 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25388101 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25388061 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25388127 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25388107 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25388064 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25388164 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25388270 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25388204 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25388073 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25388093 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25388191 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25388252 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25388200 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25388266 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25388208 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25388202 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25388180 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25388062 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25388090 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25388203 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25388124 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25388232 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25388096 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25388126 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25388080 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25388212 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25388128 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25388209 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P06196750 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25388268 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P06338782 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25388240 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25388246 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25388252 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P06052807 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P06405738 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25388243 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P06559898 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P06405731 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25388265 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25388270 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25388266 |
- |
- |