Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00493605_m1
          See other PRSS40 GE Assays ›
          Gene Symbol:
          Prss40
          Gene Name:
          protease, serine 40
          Gene Aliases:
          Tesp2
          Chromosome Location:
          Chr.1: 34536515 - 34560993 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00493605_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Prss40
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_009356.2 NP_033382.2 3 - 4 652 80
          GenBank mRNA AB008911.1 - 3 - 4 635 80
          AK006524.1 - 1 - 2 262 80
          AK132909.1 - 3 - 4 682 80
          BC145989.1 - 3 - 4 667 40131292 80

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0100002154.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Protease Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Prss40

          Panther Classification:

          Molecular Function -

          serine protease

          Gene Ontology Categories:

          Function(s) Process(es)

          proteolysis
          serine-type endopeptidase activity
          peptidase activity
          serine-type peptidase activity
          hydrolase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P48480 AK006524.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P48480 NM_009356.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P48480 BC145989.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P48480 AK006524.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P48480 NM_009356.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P48480 BC145989.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2059030 AK132909.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 352014 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 556639 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 309814 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 473818 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 754921 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 322586 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P106148 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2059030 AK132909.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P48480 AK006524.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P48480 NM_009356.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P48480 BC145989.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P48480 AK006524.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P48480 NM_009356.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P48480 BC145989.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P197805 AB008911.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20139475 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20139467 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20139468 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20139477 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20139454 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20139466 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20139472 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20139442 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20139438 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20139445 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20139456 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20139440 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20139470 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20139469 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20139444 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20139451 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20139452 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20139465 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20139441 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20139450 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20139479 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20139439 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20139478 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20139459 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20139463 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20139461 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P106148 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04059441 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04059418 - -
          Whole Mouse Genome G4122A A_52_P48480 AK006524.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P48480 NM_009356.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P48480 BC145989.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P48480 AK006524.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P48480 NM_009356.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P48480 BC145989.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P197805 AB008911.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P102462 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P117084 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P106148 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00546963 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00546962 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00546961 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00219332 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20139442 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20139407 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20139456 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20139438 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20139444 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20139413 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20139389 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20139439 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20139482 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20139480 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20139451 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20139382 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20139369 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20139363 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20139403 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20139450 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20139440 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20139459 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20139360 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20139370 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20139412 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20139414 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20139417 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20139385 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20139368 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20139401 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20139364 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20139437 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20139409 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20139376 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20139395 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20139486 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20139400 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20139398 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20139441 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20139411 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20139361 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20139392 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20139373 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20139463 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20139483 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20139465 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20139475 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20139484 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20139435 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20139404 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20139454 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20139468 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20139477 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20139406 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20139383 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00546963 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20139437 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20139440 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20139468 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00566914 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20139449 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00572974 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00500556 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20139463 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00301013 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00546961 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20139461 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20139435 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20139479 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20139438 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20139400 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20139444 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00804932 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20139411 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20139451 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00299646 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20139445 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20139456 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00572973 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00606230 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20139459 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20139414 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00718407 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00219332 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20139442 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20139465 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20139401 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00500555 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20139450 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00180262 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00769455 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20139417 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00500559 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00769449 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00606228 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20139409 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20139405 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00769448 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20139457 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20139439 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10353791 - -
          Affymetrix Mouse Gene 1.0 ST Array 10353788 - -
          Affymetrix Mouse Gene 1.0 ST Array 10353787 - -
          Affymetrix Mouse Gene 1.0 ST Array 10353789 - -
          Affymetrix Mouse Gene 1.0 ST Array 10353790 - -
          Affymetrix HT MG-430 PM Array Plate 1432486_PM_a_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10353786 - -
          Mouse Genome 430 2.0 Array 1432486_PM_a_at - -
          Mouse Genome 430 2.0 Array 1432486_a_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 21756_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 21756_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0100017272.mm.1 NM_009356.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100017272.mm.1 NM_009356.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0100017272.mm.1 TC0100002154.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100017271.mm.1 NM_009356.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100017271.mm.1 NM_009356.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0100017271.mm.1 TC0100002154.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100017275.mm.1 NM_009356.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100017275.mm.1 NM_009356.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0100017275.mm.1 TC0100002154.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100017273.mm.1 NM_009356.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100017273.mm.1 NM_009356.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0100017273.mm.1 TC0100002154.mm.1 MTA TC
          Mouse Expression Set 430 1432486_a_at - -
          Mouse Expression Set 430 1432486_PM_a_at - -
          Murine Genome U74v2 Set 94741_at - -
          Murine Genome U74v2 Set 94741_PM_at - -
          Murine Genome U74v2 Set 167738_PM_f_at - -
          Murine Genome U74v2 Set 167738_f_at - -
          Mouse Genome 430A 2.0 Array 1432486_a_at - -
          Mouse Genome 430A 2.0 Array 1432486_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1432486_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1432486_PM_a_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 509267 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 2340403 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002340403 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2771861 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_3161847 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_3161847 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2771861 - -
          mouse-6 v1.1 (Illumina) scl16882.5.1_130-S - -
          mouse-6 v1.1 (Illumina) GI_6678294-S - -
          Illumina Sentrix mouse-6.v1 bead array 2340403 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl16882.5.1_130-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2340403 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2771861 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2771861 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline