Search
Search

Ift88| Gene Symbol: Ift88 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009376.2 | NP_033402.2 | 13 - 14 | 1242 | 76 | |
| GenBank mRNA | AK135110.1 | - | 13 - 14 | 1237 | 76 | |
| AK168395.1 | - | 12 - 13 | 1151 | 76 | ||
| BC012250.1 | - | 13 - 14 | 1222 | 4019334 | 76 | |
| L31959.1 | - | 13 - 14 | 1187 | 76 | ||
Set Membership: |
|