Search
Search
| Gene Symbol: Thy1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009382.3 | NP_033408.1 | 2 - 3 | 155 | 68 | |
| GenBank mRNA | AY445633.1 | - | 1 - 2 | 37 | 68 | |
| BC054436.1 | - | 1 - 2 | 66 | 6312210 | 68 | |
| CX123560.1 | - | 2 - 3 | 155 | 68 | ||
Set Membership: |
|