Search
Search
| Gene Symbol: Ttc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009441.2 | NP_033467.2 | 15 - 16 | 1434 | 98 | |
| GenBank mRNA | AB008516.1 | - | 14 - 15 | 1311 | 98 | |
| AK053765.1 | - | 15 - 16 | 1538 | 98 | ||
| AK084201.1 | - | 5 - 6 | 469 | 98 | ||
| AK088273.1 | - | 14 - 15 | 1381 | 98 | ||
| AK165850.1 | - | 4 - 5 | 342 | 98 | ||
| AK166120.1 | - | 5 - 6 | 374 | 98 | ||
| AK166164.1 | - | 6 - 7 | 523 | 98 | ||
| AK220495.1 | - | 15 - 16 | 1476 | 98 | ||
| BC019173.1 | - | 15 - 16 | 1403 | 98 | ||
| BC037654.1 | - | 14 - 15 | 1336 | 98 | ||
| BC057207.1 | - | 14 - 15 | 1338 | 98 | ||
Set Membership: |
|