Search
Search
| Gene Symbol: Vil1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009509.2 | NP_033535.2 | 9 - 10 | 1009 | 55 | |
| GenBank mRNA | AK027908.1 | - | 9 - 10 | 1006 | 55 | |
| BC015267.1 | - | 9 - 10 | 985 | 4236751 | 55 | |
| BC117875.2 | - | 9 - 10 | 974 | 40095718 | 55 | |
| BC145671.1 | - | 9 - 10 | 976 | 40130974 | 55 | |
| M98454.1 | - | 9 - 10 | 999 | 55 | ||
Set Membership: |
|