Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00494304_m1
          See other ZFP105 GE Assays ›
          Gene Symbol:
          Zfp105
          Gene Name:
          zinc finger protein 105
          Gene Aliases:
          AW557864
          Chromosome Location:
          Chr.9: 122923040 - 122931042 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00494304_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Zfp105
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_009544.2 NP_033570.2 3 - 4 569 108
          GenBank mRNA AF045565.1 - 3 - 4 559 108
          AK147198.1 - 3 - 4 108
          BC052165.1 - 3 - 4 569 6515711 108

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0900001650.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Zfp105

          Panther Classification:

          Molecular Function -

          C2H2 zinc finger transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of transcription, DNA-templated
          transcription factor activity, sequence-specific DNA binding
          sequence-specific DNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26892102 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26892103 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26892104 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26892101 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P415225 AF045565.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P415225 AF045565.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 784328 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 618386 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 465206 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 627469 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 508916 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P149707 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P415225 AF045565.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P415225 AF045565.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26892117 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26892080 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26892119 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26892101 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26892077 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26892083 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26892122 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26892091 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26892110 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26892112 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26892099 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26892103 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26892097 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26892089 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26892106 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26892095 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26892084 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26892113 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26892082 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26892093 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26892085 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26892087 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26892081 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26892121 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26892094 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P149707 - -
          Whole Mouse Genome G4122A A_51_P415225 AF045565.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P415225 AF045565.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P137975 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P149707 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08172133 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07856503 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07852334 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07852336 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07661172 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26892101 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26892104 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26892102 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26892103 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26892081 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26892143 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26892050 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26892160 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26892062 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26892112 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26892083 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26892144 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26892165 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26892148 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26892048 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26892121 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26892119 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26892106 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26892068 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26892184 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26892158 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26892126 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26892099 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26892056 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26892130 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26892101 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26892052 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26892054 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26892123 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26892151 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26892138 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26892170 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26892139 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26892059 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26892094 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26892156 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26892177 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26892088 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26892150 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26892153 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26892155 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26892182 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26892095 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26892178 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26892084 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26892103 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26892096 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26892082 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26892174 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26892167 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26892117 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26892135 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26892124 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26892171 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26892189 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26892127 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26892122 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26892163 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26892142 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26892141 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07673349 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26892115 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07990570 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07916042 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07560641 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07852325 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26892123 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26892092 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26892091 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26892087 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26892072 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26892112 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26892068 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26892121 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26892106 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26892129 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26892119 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07658285 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26892133 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26892079 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26892077 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07667988 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26892130 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26892082 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26892134 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26892126 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26892081 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07673348 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26892141 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07948674 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07916037 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07916043 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26892117 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26892096 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26892093 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26892144 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26892135 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26892099 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07658284 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26892127 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07860905 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26892084 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07492498 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26892095 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26892094 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26892110 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10590493 - -
          Affymetrix Mouse Gene 1.0 ST Array 10590492 - -
          Affymetrix Mouse Gene 1.0 ST Array 10590491 - -
          Affymetrix Mouse Gene 1.0 ST Array 10590490 - -
          Affymetrix HT MG-430 PM Array Plate 1460252_PM_s_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10590489 - -
          Mouse Genome 430 2.0 Array 1460252_PM_s_at - -
          Mouse Genome 430 2.0 Array 1460252_s_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 22646_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 22646_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0900013847.mm.1 NM_009544.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900013847.mm.1 NM_009544.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0900013847.mm.1 TC0900001650.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900013848.mm.1 NM_009544.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900013848.mm.1 NM_009544.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0900013848.mm.1 TC0900001650.mm.1 MTA TC
          Mouse Expression Set 430 1460252_s_at - -
          Mouse Expression Set 430 1460252_PM_s_at - -
          Murine Genome U74v2 Set 95318_at - -
          Murine Genome U74v2 Set 95318_PM_at - -
          Mouse Genome 430A 2.0 Array 1460252_PM_s_at - -
          Mouse Genome 430A 2.0 Array 1460252_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1460252_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1460252_PM_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 488282 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 670156 - -
          Illumina mouse-6 v1.0 expression beadchip 670156 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000670156 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1245099 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1245099 - -
          mouse-6 v1.1 (Illumina) scl36301.4_348-S - -
          Illumina Sentrix mouse-6.v1 bead array 670156 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl36301.4_348-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 670156 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1245099 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1245099 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline