Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00494529_m1
          See other DPM2 GE Assays ›
          Gene Symbol:
          Dpm2
          Gene Name:
          dolichol-phosphate (beta-D) mannosyltransferase 2
          Gene Aliases:
          AW557993, R75484
          Chromosome Location:
          Chr.2: 32570858 - 32573571 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00494529_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Dpm2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_010073.2 NP_034203.1 2 - 3 144 93
          GenBank mRNA AB013360.1 - 2 - 3 116 93
          AK002468.1 - 2 - 3 128 93
          AK004241.1 - 2 - 3 119 93
          BC008256.1 - 2 - 3 144 2648752 93

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200005469.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Dpm2

          Gene Ontology Categories:

          Function(s) Process(es)

          GPI anchor biosynthetic process
          macromolecule biosynthetic process
          dolichol metabolic process
          regulation of protein stability
          protein O-linked mannosylation
          regulation of catalytic activity
          mannosylation
          dolichyl-phosphate beta-D-mannosyltransferase activity
          protein binding
          enzyme regulator activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P80444 NM_010073.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P80444 AK002468.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P80444 AK004241.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P80444 BC008256.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P80444 NM_010073.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P80444 AK002468.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P80444 AK004241.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P80444 BC008256.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 677806 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P166919 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P80444 NM_010073.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P80444 AK002468.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P80444 AK004241.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P80444 BC008256.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P80444 NM_010073.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P80444 AK002468.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P80444 AK004241.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P80444 BC008256.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21116128 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21116142 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21116121 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21116123 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21116120 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21116116 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21116114 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21116141 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21116126 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21116136 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21116130 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21116118 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21116132 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21116143 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21116138 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21116117 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21116131 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21116122 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21116133 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21116125 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21116135 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P166919 - -
          Whole Mouse Genome G4122A A_52_P80444 NM_010073.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P80444 AK002468.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P80444 AK004241.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P80444 BC008256.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P80444 NM_010073.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P80444 AK002468.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P80444 AK004241.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P80444 BC008256.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P104071 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P166919 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01804149 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02255658 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02255660 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02255656 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02255652 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21116132 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21116125 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21116178 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21116170 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21116203 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21116200 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21116180 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21116154 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21116123 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21116136 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21116141 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21116166 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21116117 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21116114 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21116216 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21116201 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21116214 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21116145 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21116135 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21116162 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21116176 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21116217 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21116138 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21116173 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21116211 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21116157 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21116128 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21116155 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21116171 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21116167 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21116131 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21116163 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21116130 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21116160 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21116159 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21116116 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21116122 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21116198 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21116190 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21116126 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21116164 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21116206 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21116197 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21116133 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21116194 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21116161 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21116148 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21116118 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21116168 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21116208 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21116183 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21116188 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21116150 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21116182 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21116120 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21116142 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02255655 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21116122 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21116132 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21116130 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01804128 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21116137 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21116114 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21116125 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01941994 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21116136 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01942000 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21116131 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02255652 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02255659 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21116142 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01892910 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01942003 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21116141 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21116118 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21116117 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10471442 - -
          Affymetrix Mouse Gene 1.0 ST Array 10471441 - -
          Affymetrix Mouse Gene 1.0 ST Array 10471440 - -
          Affymetrix Mouse Gene 1.0 ST Array 10471439 - -
          Affymetrix HT MG-430 PM Array Plate 1415675_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10471438 - -
          Mouse Genome 430 2.0 Array 1415675_PM_at - -
          Mouse Genome 430 2.0 Array 1415675_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 13481_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 13481_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0200005269.mm.1 NM_010073.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200005269.mm.1 NM_010073.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200005269.mm.1 TC0200005469.mm.1 MTA TC
          Mouse Expression Set 430 1415675_at - -
          Mouse Expression Set 430 1415675_PM_at - -
          Mouse Expression Set 430 1415675_at - -
          Mouse Expression Set 430 1415675_PM_at - -
          Murine Genome U74v2 Set 94052_at - -
          Murine Genome U74v2 Set 94052_PM_at - -
          Murine Genome U74v2 Set 161349_PM_f_at - -
          Murine Genome U74v2 Set 161349_f_at - -
          Mouse Genome 430A 2.0 Array 1415675_at - -
          Mouse Genome 430A 2.0 Array 1415675_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1415675_at - -
          Affymetrix HT Mouse Genome 430A Array 1415675_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1415675_at - -
          Affymetrix HT Mouse Genome 430B Array 1415675_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 409810 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 106520739 - -
          Illumina mouse-6 v1.0 expression beadchip 3610253 - -
          Illumina mouse-6 v1.0 expression beadchip 106520739 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003610253 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2743683 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2743683 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1232765 - -
          mouse-6 v1.1 (Illumina) scl013481.4_295-S - -
          Illumina Sentrix mouse-6.v1 bead array 3610253 - -
          Illumina Sentrix mouse-6.v1 bead array 106520739 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl013481.4_295-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 106520739 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3610253 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2930095 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2930095 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2583343 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl013481.4_295-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline