Search
Search
| Gene Symbol: Gli1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010296.2 | NP_034426.2 | 11 - 12 | 1476 | 83 | |
| GenBank mRNA | AB025922.1 | - | 11 - 12 | 1486 | 83 | |
| AF026305.1 | - | 11 - 12 | 1476 | 83 | ||
| BC131650.1 | - | 10 - 11 | 1454 | 40110429 | 83 | |
Set Membership: |
|