Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 我們的服務對象
    • 生物技術
    • 生物製藥
    • CDMO
    • 實驗室診斷
    • 工業與應用科學
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00494686_g1
          See other GNG3 GE Assays ›
          Gene Symbol:
          Gng3
          Gene Name:
          guanine nucleotide binding protein (G protein), gamma 3
          Gene Aliases:
          -
          Chromosome Location:
          Chr.19: 8836929 - 8839246 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00494686_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gng3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_010316.3 NP_034446.1 2 - 3 360 78
          GenBank mRNA AK004459.1 - 2 - 3 325 78
          AK032646.1 - 2 - 3 308 78
          BC029680.1 - 2 - 3 244 5365311 78
          U38497.1 - 1 - 2 67 78

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1900001072.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Gng3

          Panther Classification:

          Molecular Function -

          heterotrimeric G-protein

          Gene Ontology Categories:

          Function(s) Process(es)

          signal transduction
          G-protein coupled receptor signaling pathway
          positive regulation of cytosolic calcium ion concentration
          G-protein coupled receptor binding
          GTPase activity
          signal transducer activity
          type 1 angiotensin receptor binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P391291 AK032646.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P391291 BC029680.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P391291 NM_010316.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P391291 AK004459.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P391291 AK032646.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P391291 BC029680.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P391291 NM_010316.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P391291 AK004459.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 680622 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 369787 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P130276 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P391291 AK032646.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P391291 BC029680.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P391291 NM_010316.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P391291 AK004459.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P391291 AK032646.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P391291 BC029680.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P391291 NM_010316.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P391291 AK004459.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P340665 AK032646.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P340665 BC029680.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P340665 NM_010316.3 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P340665 AK004459.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P391291 AK032646.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P391291 BC029680.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P391291 NM_010316.3 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P391291 AK004459.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P391291 AK032646.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P391291 BC029680.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P391291 NM_010316.3 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P391291 AK004459.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P391291 AK032646.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P391291 BC029680.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P391291 NM_010316.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P391291 AK004459.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P391291 AK032646.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P391291 BC029680.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P391291 NM_010316.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P391291 AK004459.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P340665 AK032646.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P340665 BC029680.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P340665 NM_010316.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P340665 AK004459.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P134230 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P100417 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14305318 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14305322 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14287182 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14305312 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14287178 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14241182 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14305315 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14305311 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14305314 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14287184 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14241181 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10465825 - -
          Affymetrix Mouse Gene 1.0 ST Array 10465822 - -
          Affymetrix Mouse Gene 1.0 ST Array 10465821 - -
          Affymetrix Mouse Gene 1.0 ST Array 10465824 - -
          Affymetrix Mouse Gene 1.0 ST Array 10465823 - -
          Affymetrix HT MG-430 PM Array Plate 1417428_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10465820 - -
          Mouse Genome 430 2.0 Array 1417428_PM_at - -
          Mouse Genome 430 2.0 Array 1417428_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 14704_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 14704_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1900010057.mm.1 NM_010316.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900010057.mm.1 NM_010316.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1900010057.mm.1 TC1900001072.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900010058.mm.1 NM_010316.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900010058.mm.1 NM_010316.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1900010058.mm.1 TC1900001072.mm.1 MTA TC
          Mouse Expression Set 430 1417428_PM_at - -
          Mouse Expression Set 430 1417428_at - -
          Murine Genome U74v2 Set 94986_PM_at - -
          Murine Genome U74v2 Set 94986_at - -
          Mouse Genome 430A 2.0 Array 1417428_PM_at - -
          Mouse Genome 430A 2.0 Array 1417428_at - -
          Affymetrix HT Mouse Genome 430A Array 1417428_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1417428_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 614586 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 6980397 - -
          Illumina mouse-6 v1.0 expression beadchip 6980397 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006980397 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1253137 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1253137 - -
          mouse-6 v1.1 (Illumina) scl52774.3_213-S - -
          Illumina Sentrix mouse-6.v1 bead array 6980397 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl52774.3_213-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6980397 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1253137 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1253137 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl52774.3_213-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline