Search
Search

Kifc2| Gene Symbol: Kifc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010630.2 | NP_034760.2 | 12 - 13 | 1413 | 81 | |
| GenBank mRNA | AK144304.1 | - | 13 - 14 | 1572 | 81 | |
| D49545.1 | - | 12 - 13 | 1489 | 81 | ||
| U92949.1 | - | 12 - 13 | 1403 | 81 | ||
Set Membership: |
|