Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00495258_m1
          See other LALBA GE Assays ›
          Gene Symbol:
          Lalba
          Gene Name:
          lactalbumin, alpha
          Gene Aliases:
          AW208827
          Chromosome Location:
          Chr.15: 98480400 - 98482934 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00495258_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Lalba
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_010679.1 NP_034809.1 3 - 4 382 65
          GenBank mRNA BC006922.1 - 3 - 4 394 3601746 65
          BC069916.1 - 3 - 4 382 3670536 65
          M80909.1 - 3 - 4 382 65

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1500002175.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Apoptosis 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Lalba

          Gene Ontology Categories:

          Function(s) Process(es)

          lactose biosynthetic process
          lactose synthase activity
          calcium ion binding
          protein binding
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2150540 BC006922.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2150540 BC069916.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2150540 M80909.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2150540 NM_010679.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2150540 BC044869.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 495737 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P146856 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2150540 BC006922.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2150540 BC069916.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2150540 M80909.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2150540 NM_010679.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2150540 BC044869.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P156232 BC006922.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P156232 BC069916.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P156232 M80909.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P156232 NM_010679.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30474776 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30474779 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30474801 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30474794 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30474773 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30474757 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30474762 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30474791 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30474781 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30474784 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30474758 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30474768 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30474800 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30474771 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30474774 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30474785 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30474778 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30474755 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30474765 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30474792 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30474796 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30474799 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30474766 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30474803 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30474789 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30474790 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30474763 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30474783 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30474770 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30474775 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30474788 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30474760 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P146856 - -
          Whole Mouse Genome G4122A A_51_P156232 BC006922.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P156232 BC069916.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P156232 M80909.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P156232 NM_010679.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P138223 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P108265 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P146856 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12365182 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12365185 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12365184 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12365188 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12365181 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12123620 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30474789 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30474708 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30474728 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30474725 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30474814 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30474788 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30474763 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30474824 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30474757 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30474702 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30474779 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30474742 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30474791 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30474741 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30474783 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30474726 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30474738 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30474812 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30474768 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30474706 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30474818 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30474771 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30474816 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30474796 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30474713 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30474703 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30474711 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30474766 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30474760 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30474803 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30474758 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30474794 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30474755 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30474731 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30474770 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30474730 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30474744 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30474765 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30474776 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30474723 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30474747 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30474799 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30474746 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30474717 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30474785 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30474790 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30474784 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30474737 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30474778 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30474774 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30474720 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30474735 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30474810 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30474807 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30474725 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30474784 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30474763 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30474758 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30474702 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30474753 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12155938 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30474785 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30474791 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30474798 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30474730 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30474760 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30474794 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12150368 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30474796 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11986303 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30474790 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30474731 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30474762 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30474713 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12155943 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30474720 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12010949 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30474793 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30474746 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12359314 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12365182 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30474781 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30474774 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30474804 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30474706 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30474771 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30474755 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30474754 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12210239 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30474744 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30474708 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30474750 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30474788 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12113997 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30474704 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30474765 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30474748 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30474738 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12306969 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12063042 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12365188 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12010952 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12306965 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30474792 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12124751 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30474773 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30474737 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12124744 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30474741 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30474776 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12210238 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30474806 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12155949 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30474803 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30474783 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30474728 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12123616 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30474768 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30474735 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30474703 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30474717 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12155944 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30474723 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10432160 - -
          Affymetrix Mouse Gene 1.0 ST Array 10432159 - -
          Affymetrix Mouse Gene 1.0 ST Array 10432161 - -
          Affymetrix Mouse Gene 1.0 ST Array 10432158 - -
          Affymetrix HT MG-430 PM Array Plate 1418363_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10432157 - -
          Mouse Genome 430 2.0 Array 1418363_at - -
          Mouse Genome 430 2.0 Array 1418363_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 16770_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 16770_at - -
          Affymetrix Mouse Clariom S Array PSR1500017633.mm.1 NM_010679.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500017633.mm.1 NM_010679.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500017633.mm.1 TC1500002175.mm.1 MTA TC
          Mouse Expression Set 430 1418363_PM_at - -
          Mouse Expression Set 430 1418363_at - -
          Murine Genome U74v2 Set 92994_PM_at - -
          Murine Genome U74v2 Set 92994_at - -
          Mouse Genome 430A 2.0 Array 1418363_at - -
          Mouse Genome 430A 2.0 Array 1418363_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1418363_at - -
          Affymetrix HT Mouse Genome 430A Array 1418363_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 352178 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 6940685 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006940685 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2676106 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2676106 - -
          mouse-6 v1.1 (Illumina) scl46770.4.1_1-S - -
          Illumina Sentrix mouse-6.v1 bead array 6940685 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl46770.4.1_1-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6940685 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2676107 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2676107 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline