Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00495384_m1
          See other LOX GE Assays ›
          Gene Symbol:
          Lox
          Gene Name:
          lysyl oxidase
          Gene Aliases:
          AI893619, TSC-160, rrg
          Chromosome Location:
          Chr.18: 52516060 - 52529867 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00495384_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Lox
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001286181.1 NP_001273110.1 3 - 4 840 72
          NM_001286182.1 NP_001273111.1 3 - 4 797 72
          NM_010728.3 NP_034858.2 2 - 3 1217 72
          GenBank mRNA AK135213.1 - 2 - 3 348 72
          AK141995.1 - 2 - 3 1103 72
          AK159199.1 - 2 - 3 1020 72
          AK159241.1 - 2 - 3 996 72
          AK159263.1 - 2 - 3 1057 72
          AK159368.1 - 2 - 3 979 72
          AK159682.1 - 2 - 3 930 72
          AK159796.1 - 2 - 3 991 72
          AK160096.1 - 2 - 3 991 72
          AK164634.1 - 3 - 4 798 72
          AK166279.1 - 2 - 3 925 72
          AK166386.1 - 1061 72
          AK166442.1 - 1217 72
          BC018439.1 - 2 - 3 1068 2655752 72
          M65142.1 - 2 - 3 985 72

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1800001334.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Lox

          Panther Classification:

          Molecular Function -

          oxidase

          Gene Ontology Categories:

          Function(s) Process(es)

          blood vessel development
          response to hormone
          collagen fibril organization
          lung development
          wound healing
          elastic fiber assembly
          response to steroid hormone
          oxidation-reduction process
          protein-lysine 6-oxidase activity
          copper ion binding
          protein binding
          oxidoreductase activity
          oxidoreductase activity, acting on the CH-NH2 group of donors, oxygen as acceptor
          metal ion binding

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1800009829.mm.1 NM_001286181.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800009829.mm.1 NM_010728.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1800009829.mm.1 TC1800001334.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1800009826.mm.1 NM_001286181.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800009826.mm.1 NM_001286182.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800009826.mm.1 NM_010728.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1800009826.mm.1 TC1800001334.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1800009827.mm.1 NM_001286181.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800009827.mm.1 NM_001286182.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800009827.mm.1 NM_010728.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1800009827.mm.1 TC1800001334.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1800009828.mm.1 NM_010728.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1800009828.mm.1 TC1800001334.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1800009832.mm.1 NM_010728.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1800009832.mm.1 TC1800001334.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1800009825.mm.1 NM_001286181.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800009825.mm.1 NM_001286182.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800009825.mm.1 NM_010728.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1800009825.mm.1 TC1800001334.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1800009830.mm.1 NM_001286181.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800009830.mm.1 NM_010728.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1800009830.mm.1 TC1800001334.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1800009833.mm.1 NM_001286182.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800009833.mm.1 TC1800001334.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 917511 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-4lm45:80/100.66.75.98:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0