Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00495657_m1
          See other CLEC3B GE Assays ›
          Gene Symbol:
          Clec3b
          Gene Name:
          C-type lectin domain family 3, member b
          Gene Aliases:
          Tna
          Chromosome Location:
          Chr.9: 123150946 - 123157432 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00495657_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Clec3b
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_011606.2 NP_035736.2 2 - 3 306 74
          GenBank mRNA BC035043.1 - 2 - 3 258 1313548 74
          X79199.1 - 2 - 3 281 74

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0900001658.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Clec3b

          Panther Classification:

          Molecular Function -

          intercellular signal molecule

          Gene Ontology Categories:

          Function(s) Process(es)

          skeletal system development
          positive regulation of plasminogen activation
          bone mineralization
          calcium ion binding
          protein binding
          heparin binding
          carbohydrate binding
          kringle domain binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26893460 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26893461 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P486239 X79199.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P486239 X79199.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 172340 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 7137 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 235613 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 9190 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 269761 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P100165 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P486239 X79199.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P486239 X79199.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P486239 X79199.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P486239 X79199.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26893399 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26893390 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26893420 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26893422 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26893397 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26893423 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26893404 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26893418 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26893387 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26893392 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26893432 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26893417 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26893434 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26893428 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26893426 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26893435 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26893402 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26893419 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26893436 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26893415 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26893391 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26893393 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26893411 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26893394 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26893425 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26893424 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26893398 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26893413 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26893430 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P100165 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P02050944 - -
          Whole Mouse Genome G4122A A_51_P486239 X79199.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P486239 X79199.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P113189 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P100165 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07917969 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07917971 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07917996 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07917986 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07917992 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07917974 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07917990 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26893461 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26893460 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26893415 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26893419 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26893467 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26893456 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26893448 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26893413 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26893459 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26893502 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26893473 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26893420 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26893435 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26893475 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26893484 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26893461 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26893394 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26893429 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26893390 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26893437 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26893422 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26893398 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26893481 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26893454 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26893441 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26893411 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26893402 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26893428 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26893391 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26893500 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26893426 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26893468 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26893505 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26893462 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26893434 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26893492 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26893451 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26893485 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26893489 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26893440 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26893495 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26893393 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26893430 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26893472 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26893432 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26893479 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26893486 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26893400 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26893477 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26893487 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26893444 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26893417 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26893423 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26893436 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26893470 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26893404 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26893497 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26893474 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26893452 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26893478 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26893483 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26893447 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26893397 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26893444 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26893417 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26893413 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26893420 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26893428 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26893386 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26893400 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26893452 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26893462 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26893432 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07799615 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26893440 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26893454 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26893397 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26893425 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26893402 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26893426 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26893390 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26893411 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07799620 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26893459 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26893434 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26893415 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07978099 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26893430 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26893423 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26893429 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26893465 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26893394 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26893449 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07480953 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26893398 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26893391 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26893447 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26893399 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26893464 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26893451 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26893468 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07830969 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26893446 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26893404 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26893457 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26893461 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26893410 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26893422 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07799617 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07830974 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26893437 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08011879 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10590561 - -
          Affymetrix Mouse Gene 1.0 ST Array 10590560 - -
          Affymetrix Mouse Gene 1.0 ST Array 10590562 - -
          Affymetrix HT MG-430 PM Array Plate 1449466_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10590558 - -
          Mouse Genome 430 2.0 Array 1449466_PM_at - -
          Mouse Genome 430 2.0 Array 1449466_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 21922_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 21922_at - -
          Affymetrix Mouse Clariom S Array PSR0900013946.mm.1 NM_011606.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900013946.mm.1 NM_011606.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0900013946.mm.1 TC0900001658.mm.1 MTA TC
          Mouse Expression Set 430 1449466_at - -
          Mouse Expression Set 430 1449466_PM_at - -
          Murine Genome U74v2 Set 92224_at - -
          Murine Genome U74v2 Set 92224_PM_at - -
          Mouse Genome 430A 2.0 Array 1449466_at - -
          Mouse Genome 430A 2.0 Array 1449466_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1449466_at - -
          Affymetrix HT Mouse Genome 430A Array 1449466_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 554974 - -
          Array Name Probe IDs Targets Target Type
          MouseRef-8 v2 Expression BeadChip ILMN_2845778 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2845778 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline