Search
Search
| Gene Symbol: Uba3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF077330.1 | - | 11 - 12 | 938 | 84 | |
| AK032514.1 | - | 12 - 13 | 1172 | 84 | ||
| AK048148.1 | - | 12 - 13 | 970 | 84 | ||
| AK159381.1 | - | 12 - 13 | 1058 | 84 | ||
| AK166655.1 | - | 11 - 12 | 928 | 84 | ||
| AK168859.1 | - | 12 - 13 | 973 | 84 | ||
| AY029181.1 | - | 11 - 12 | 938 | 84 | ||
| BC002002.1 | - | 12 - 13 | 958 | 3590282 | 84 | |
| BC080776.1 | - | 11 - 12 | 891 | 84 | ||
| CF928750.1 | - | 1 - 2 | 71 | 84 | ||
Set Membership: |
|