Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00495912_m1
          See other UFD1L GE Assays ›
          Gene Symbol:
          Ufd1l
          Gene Name:
          ubiquitin fusion degradation 1 like
          Gene Aliases:
          Ufd1
          Chromosome Location:
          Chr.16: 18811801 - 18835261 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00495912_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ufd1l
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_011672.4 NP_035802.3 3 - 4 289 126
          GenBank mRNA AK010461.1 - 3 - 4 258 126
          AK088823.1 - 3 - 4 258 126
          AK147138.1 - 3 - 4 272 126
          AK169062.1 - 3 - 4 271 126
          BC006630.1 - 3 - 4 246 3708320 126
          CJ085208.1 - 3 - 4 289 126
          U64445.1 - 3 - 4 274 126

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1600000250.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ufd1l

          Gene Ontology Categories:

          Function(s) Process(es)

          ubiquitin-dependent protein catabolic process
          retrograde protein transport, ER to cytosol
          ER-associated misfolded protein catabolic process
          receptor binding
          protein complex binding
          ATPase binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P220347 U64445.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P220347 AK088823.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P220347 AK147138.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P220347 NM_011672.4 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P220347 NR_028403.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P220347 U64445.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P220347 AK088823.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P220347 AK147138.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P220347 NM_011672.4 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P220347 NR_028403.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2047509 BC006630.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2047509 AK012543.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2047509 AK088823.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2047509 AK147138.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2047509 AK169062.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 347034 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 317947 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 676996 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 498713 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 41951 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 854105 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 138596 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 510022 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 906354 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 139432 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 842926 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 113083 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 713676 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 546267 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 489977 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 126492 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 160531 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P103079 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P155387 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2047509 BC006630.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2047509 AK012543.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2047509 AK088823.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2047509 AK147138.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2047509 AK169062.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P220347 U64445.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P220347 AK088823.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P220347 AK147138.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P220347 NM_011672.4 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P220347 NR_028403.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P220347 U64445.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P220347 AK088823.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P220347 AK147138.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P220347 NM_011672.4 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P220347 NR_028403.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P220347 U64445.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P220347 AK088823.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P220347 AK147138.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P220347 NM_011672.4 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P220347 NR_028403.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P220347 U64445.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P220347 AK088823.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P220347 AK147138.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P220347 NM_011672.4 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P220347 NR_028403.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P328963 U64445.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P123858 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13006414 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13006402 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13006415 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13006429 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10434245 - -
          Affymetrix Mouse Gene 1.0 ST Array 10434247 - -
          Affymetrix Mouse Gene 1.0 ST Array 10434246 - -
          Affymetrix Mouse Gene 1.0 ST Array 10434243 - -
          Affymetrix Mouse Gene 1.0 ST Array 10434244 - -
          Affymetrix Mouse Gene 1.0 ST Array 10434234 - -
          Affymetrix Mouse Gene 1.0 ST Array 10434242 - -
          Affymetrix Mouse Gene 1.0 ST Array 10434240 - -
          Affymetrix Mouse Gene 1.0 ST Array 10434238 - -
          Affymetrix Mouse Gene 1.0 ST Array 10434239 - -
          Affymetrix Mouse Gene 1.0 ST Array 10434235 - -
          Affymetrix Mouse Gene 1.0 ST Array 10434237 - -
          Affymetrix Mouse Gene 1.0 ST Array 10434241 - -
          Affymetrix Mouse Gene 1.0 ST Array 10434236 - -
          Affymetrix HT MG-430 PM Array Plate 1432367_PM_a_at - -
          Affymetrix HT MG-430 PM Array Plate 1418087_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10434233 - -
          Mouse Genome 430 2.0 Array 1418087_at - -
          Mouse Genome 430 2.0 Array 1432367_a_at - -
          Mouse Genome 430 2.0 Array 1432367_PM_a_at - -
          Mouse Genome 430 2.0 Array 1418087_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 22230_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 22230_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1600002306.mm.1 NM_011672.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600002306.mm.1 NM_011672.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1600002306.mm.1 NR_028403.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600002306.mm.1 NR_028403.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600002306.mm.1 TC1600000250.mm.1 MTA TC
          Mouse Expression Set 430 1418087_at - -
          Mouse Expression Set 430 1432367_PM_a_at - -
          Mouse Expression Set 430 1418087_PM_at - -
          Mouse Expression Set 430 1432367_a_at - -
          Murine Genome U74v2 Set 93303_PM_at - -
          Murine Genome U74v2 Set 93303_at - -
          Mouse Genome 430A 2.0 Array 1418087_at - -
          Mouse Genome 430A 2.0 Array 1432367_PM_a_at - -
          Mouse Genome 430A 2.0 Array 1432367_a_at - -
          Mouse Genome 430A 2.0 Array 1418087_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1432367_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1418087_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1432367_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1418087_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 423140 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2443482 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2443482 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline