| Array Name |
Probe IDs |
Targets |
Target Type |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P424532 |
BC019203.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P424532 |
AJ132098.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P424532 |
BC019203.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P424532 |
AJ132098.1 |
mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
258711 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
899469 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
602403 |
AK137439.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
602403 |
BC055811.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
602403 |
NM_198610.1 |
refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
623794 |
AF150963.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
623794 |
AK030031.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
623794 |
NM_011667.2 |
refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
623794 |
X62581.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
623794 |
XM_001474599.1 |
refseq_mrna_mouse_XM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
654653 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
758404 |
AK131845.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
758404 |
AK131994.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
758404 |
AK149015.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
758404 |
AK157925.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
758404 |
AK158714.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
758404 |
BC132217.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
758404 |
BC132219.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
758404 |
L12140.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
758404 |
NM_010347.3 |
refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
758404 |
X73359.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
758404 |
X73361.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
111692 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
94565 |
- |
- |
| Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P120214 |
- |
- |
| Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P169223 |
- |
- |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P424532 |
BC019203.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P424532 |
AJ132098.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P424532 |
BC019203.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P424532 |
AJ132098.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P424532 |
BC019203.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P424532 |
AJ132098.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P424532 |
BC019203.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P424532 |
AJ132098.1 |
mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27005630 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27005656 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27005639 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27005618 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27005637 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27005622 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27005620 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27005650 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27005645 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27005619 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27005649 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27005617 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27005633 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27005642 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27005654 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27005647 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27005632 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27005626 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27005625 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27005635 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27005624 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27005616 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27005628 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27005652 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27005646 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P169223 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P07003107 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P120214 |
- |
- |
| Whole Mouse Genome G4122A |
A_51_P424532 |
BC019203.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P424532 |
AJ132098.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P424532 |
BC019203.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P424532 |
AJ132098.1 |
mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P102249 |
- |
- |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P169223 |
- |
- |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P120214 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P08853969 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P08853975 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P08437807 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P08853970 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27005696 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27005652 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27005651 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27005632 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27005636 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27005718 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27005726 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27005611 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27005630 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27005768 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27005747 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27005685 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27005616 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27005598 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27005751 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27005702 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27005689 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27005734 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27005682 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27005631 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27005760 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27005721 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27005649 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27005704 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27005755 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27005615 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27005740 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27005642 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27005709 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27005680 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27005604 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27005772 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27005672 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27005771 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27005646 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27005675 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27005665 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27005761 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27005605 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27005639 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27005599 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27005613 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27005703 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27005757 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27005763 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27005733 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27005693 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27005739 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27005659 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27005710 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27005706 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27005683 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27005724 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27005714 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27005618 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27005656 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27005626 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27005607 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27005700 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27005669 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27005765 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27005678 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27005753 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27005635 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27005695 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27005657 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27005749 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27005745 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27005743 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27005712 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27005608 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27005640 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27005596 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27005692 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27005728 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27005625 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27005645 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P08330370 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27005687 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27005620 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27005633 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P08422402 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P08292997 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27005635 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27005640 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27005682 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27005655 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27005608 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27005693 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27005616 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27005692 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27005665 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P08242635 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P08677372 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27005654 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27005625 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27005688 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27005638 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P08945996 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27005605 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27005649 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27005613 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27005669 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27005659 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27005685 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27005630 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P08583128 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27005607 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27005632 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27005661 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27005615 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27005675 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27005646 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P08255867 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27005658 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27005689 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P08853975 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27005680 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P08677367 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27005617 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27005639 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27005610 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27005618 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27005628 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27005651 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27005683 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27005642 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27005657 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27005622 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27005678 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P08948790 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27005626 |
- |
- |