Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00496067_m1
          See other ZFP54 GE Assays ›
          Gene Symbol:
          Zfp54
          Gene Name:
          zinc finger protein 54
          Gene Aliases:
          2810034D10Rik, KRAB10, Zfp-54, Zfp76, mszf83
          Chromosome Location:
          Chr.17: 21423227 - 21435640 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00496067_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Zfp54
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_011760.2 NP_035890.2 3 - 4 553 126
          GenBank mRNA AF080070.1 - 3 - 4 514 126
          AK148292.1 - 3 - 4 546 126
          BC080782.1 - 4 - 5 605 30468186 126

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1700000270.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Zfp54

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of transcription, DNA-templated
          transcription factor activity, sequence-specific DNA binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2159875 BC080782.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2159875 NM_011760.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2017075 AK148292.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2017075 BC080782.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2017075 M74236.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2017075 AF080070.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2017075 NM_011760.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 203858 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 715581 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 487207 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 905460 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 425965 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 51240 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 688569 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 306842 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 146064 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 118119 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P148756 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P162552 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P124188 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P162723 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P123957 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2017075 AK148292.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2017075 BC080782.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2017075 M74236.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2017075 AF080070.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2017075 NM_011760.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2159875 BC080782.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2159875 NM_011760.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31086002 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31085985 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31085983 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31085981 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31085990 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31085992 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31085996 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31085989 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31085993 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31085995 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31085988 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31085994 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P162723 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P123957 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08733988 - -
          Whole Mouse Genome G4122A A_51_P392717 AF080070.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P125397 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P123957 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P162723 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13382142 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13382145 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13382146 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31085993 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31085994 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31086015 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31086055 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31086045 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31086009 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31085975 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31086007 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31086025 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31086032 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31086003 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31085988 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31086014 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31085985 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31085995 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31086029 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31086019 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31086049 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31086058 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31085981 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31085979 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31085983 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31086051 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31086046 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31086016 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31086053 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31086011 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31085990 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31086010 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31085992 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31086004 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31086023 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13421490 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31085990 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31085983 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13106651 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31085988 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31086009 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13106652 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31086004 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31085985 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13363281 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31086003 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31086002 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31086021 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31086019 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13363284 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31085992 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31085999 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31085981 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31086016 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31086010 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31085993 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13382136 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13106644 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31086007 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31086013 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10442204 - -
          Affymetrix Mouse Gene 1.0 ST Array 10442202 - -
          Affymetrix Mouse Gene 1.0 ST Array 10442203 - -
          Affymetrix Mouse Gene 1.0 ST Array 10442205 - -
          Affymetrix Mouse Gene 1.0 ST Array 10442200 - -
          Affymetrix Mouse Gene 1.0 ST Array 10442201 - -
          Affymetrix HT MG-430 PM Array Plate 1454584_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1419239_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10442199 - -
          Mouse Genome 430 2.0 Array 1419239_PM_at - -
          Mouse Genome 430 2.0 Array 1419239_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 22712_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 22712_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1700001979.mm.1 TC1700000270.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700001976.mm.1 NM_011760.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700001976.mm.1 NM_011760.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700001976.mm.1 TC1700000270.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700001977.mm.1 NM_011760.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700001977.mm.1 NM_011760.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700001977.mm.1 TC1700000270.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700001978.mm.1 NM_011760.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700001978.mm.1 NM_011760.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700001978.mm.1 TC1700000270.mm.1 MTA TC
          Mouse Expression Set 430 1419239_PM_at - -
          Mouse Expression Set 430 1419239_at - -
          Murine Genome U74v2 Set 102625_f_at - -
          Murine Genome U74v2 Set 102626_PM_r_at - -
          Murine Genome U74v2 Set 102626_r_at - -
          Murine Genome U74v2 Set 102625_PM_f_at - -
          Mouse Genome 430A 2.0 Array 1419239_at - -
          Mouse Genome 430A 2.0 Array 1419239_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1454584_at - -
          Affymetrix HT Mouse Genome 430B Array 1454584_PM_at - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 1340091 - -
          Illumina mouse-6 v1.0 expression beadchip 1340091 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001340091 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2469069 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2469069 - -
          mouse-6 v1.1 (Illumina) scl022712.7_302-S - -
          Illumina Sentrix mouse-6.v1 bead array 1340091 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl022712.7_302-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1340091 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2469069 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2469069 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline