Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00496424_s1
          See other ZFP260 GE Assays ›
          Gene Symbol:
          Zfp260
          Gene Name:
          zinc finger protein 260
          Gene Aliases:
          AI480997, Ozrf1, PEX1, Zfp63, Znf260
          Chromosome Location:
          Chr.7: 30094782 - 30107622 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00496424_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Zfp260
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_011981.4 NP_036111.2 3 1029 77
          GenBank mRNA AK088177.1 - 1 551 77
          AK156219.1 - 3 919 77
          BC082570.1 - 1 355 30607787 77
          BC085180.1 - 3 1029 30639007 77
          D45210.1 - 2 841 77

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700000546.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Zfp260

          Panther Classification:

          Molecular Function -

          C2H2 zinc finger transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          regulation of transcription, DNA-templated
          multicellular organism development
          nucleic acid binding
          DNA binding
          transcription factor activity, sequence-specific DNA binding
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2027771 BC082570.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2027771 BC085180.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2027771 AK156219.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2027771 AK166377.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2027771 BC024566.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 278651 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 484907 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 36152 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 663775 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 367669 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 661368 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 528374 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 683399 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 32434 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 128925 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P164050 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2027771 BC082570.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2027771 BC085180.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2027771 AK156219.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2027771 AK166377.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2027771 BC024566.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P156781 BC082570.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P156781 BC085180.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P156781 AK156219.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P156781 AK088177.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P156781 NM_011981.4 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25029877 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25029880 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25029879 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25029881 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25029875 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25029883 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25029873 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25029882 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01492519 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P164050 - -
          Whole Mouse Genome G4122A A_51_P156781 BC082570.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P156781 BC085180.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P156781 AK156219.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P156781 AK088177.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P156781 NM_011981.4 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P649721 D45210.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P649721 BC082570.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P649721 BC085180.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P649721 AK156219.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P649721 AK088177.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P110331 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P164050 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06141027 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25029850 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25029852 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25029928 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25029908 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25029893 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25029869 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25029895 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25029923 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25029867 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25029890 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25029875 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25029911 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25029914 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25029919 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25029947 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25029846 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25029853 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25029942 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25029849 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25029883 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25029929 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25029940 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25029832 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25029938 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25029924 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25029841 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25029845 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25029877 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25029864 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25029837 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25029880 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25029933 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25029836 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25029916 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25029913 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25029856 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25029891 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25029909 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25029872 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25029847 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25029896 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25029889 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25029902 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25029955 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25029830 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25029899 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25029829 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25029900 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25029906 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25029888 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25029949 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25029921 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25029887 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25029936 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25029854 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06542091 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06096457 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25029888 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25029877 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25029893 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06392402 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25029900 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25029862 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25029872 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25029899 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25029869 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25029887 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25029895 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25029881 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25029875 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25029867 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25029902 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25029904 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25029882 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25029890 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25029880 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25029891 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25029883 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06041833 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25029889 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10551818 - -
          Affymetrix Mouse Gene 1.0 ST Array 10551821 - -
          Affymetrix Mouse Gene 1.0 ST Array 10551817 - -
          Affymetrix Mouse Gene 1.0 ST Array 10551816 - -
          Affymetrix Mouse Gene 1.0 ST Array 10551820 - -
          Affymetrix Mouse Gene 1.0 ST Array 10551819 - -
          Affymetrix HT MG-430 PM Array Plate 1419164_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1427831_PM_s_at - -
          Affymetrix HT MG-430 PM Array Plate 1419165_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1427830_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1436495_PM_s_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10551815 - -
          Mouse Genome 430 2.0 Array 1436495_PM_s_at - -
          Mouse Genome 430 2.0 Array 1419165_PM_at - -
          Mouse Genome 430 2.0 Array 1419165_at - -
          Mouse Genome 430 2.0 Array 1419164_PM_at - -
          Mouse Genome 430 2.0 Array 1427831_s_at - -
          Mouse Genome 430 2.0 Array 1427830_PM_at - -
          Mouse Genome 430 2.0 Array 1436495_s_at - -
          Mouse Genome 430 2.0 Array 1419164_at - -
          Mouse Genome 430 2.0 Array 1427831_PM_s_at - -
          Mouse Genome 430 2.0 Array 1427830_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 26466_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 26466_at - -
          Affymetrix Murine Genome 6500A Array Msa12980_at - -
          Affymetrix Mouse Clariom S Array PSR0700004686.mm.1 TC0700000546.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700004686.mm.1 NM_011981.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700004686.mm.1 NM_011981.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0700004685.mm.1 TC0700000546.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700004685.mm.1 NM_011981.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700004685.mm.1 NM_011981.4 Refseq
          Mouse Expression Set 430 1427830_PM_at - -
          Mouse Expression Set 430 1427831_s_at - -
          Mouse Expression Set 430 1419164_at - -
          Mouse Expression Set 430 1419165_PM_at - -
          Mouse Expression Set 430 1427830_at - -
          Mouse Expression Set 430 1436495_s_at - -
          Mouse Expression Set 430 1419164_PM_at - -
          Mouse Expression Set 430 1419165_at - -
          Mouse Expression Set 430 1436495_PM_s_at - -
          Mouse Expression Set 430 1427831_PM_s_at - -
          Murine Genome U74v2 Set 95801_s_at - -
          Murine Genome U74v2 Set 95801_PM_s_at - -
          Murine Genome U74v2 Set 92409_PM_at - -
          Murine Genome U74v2 Set 92409_at - -
          Mouse Genome 430A 2.0 Array 1427830_at - -
          Mouse Genome 430A 2.0 Array 1419164_PM_at - -
          Mouse Genome 430A 2.0 Array 1427831_s_at - -
          Mouse Genome 430A 2.0 Array 1427831_PM_s_at - -
          Mouse Genome 430A 2.0 Array 1419165_at - -
          Mouse Genome 430A 2.0 Array 1427830_PM_at - -
          Mouse Genome 430A 2.0 Array 1419164_at - -
          Mouse Genome 430A 2.0 Array 1436495_s_at - -
          Mouse Genome 430A 2.0 Array 1419165_PM_at - -
          Mouse Genome 430A 2.0 Array 1436495_PM_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1436495_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1419165_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1427830_at - -
          Affymetrix HT Mouse Genome 430A Array 1419164_at - -
          Affymetrix HT Mouse Genome 430A Array 1419164_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1436495_PM_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1427830_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1427831_PM_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1419165_at - -
          Affymetrix HT Mouse Genome 430A Array 1427831_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 790904 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 1990301 - -
          Illumina mouse-6 v1.0 expression beadchip 101850059 - -
          Illumina mouse-6 v1.0 expression beadchip 1990301 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001990301 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2476728 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2476728 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1247633 - -
          mouse-6 v1.1 (Illumina) scl0026466.2_245-S - -
          mouse-6 v1.1 (Illumina) scl32827.4_67-S - -
          Illumina Sentrix mouse-6.v1 bead array 101850059 - -
          Illumina Sentrix mouse-6.v1 bead array 1990301 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0026466.2_245-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 101850059 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1990301 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2441326 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1247633 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-4lm45:80/100.66.75.98:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0