Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00496525_m1
          See other DSC1 GE Assays ›
          Gene Symbol:
          Dsc1
          Gene Name:
          desmocollin 1
          Gene Aliases:
          1110020A10Rik, AI507491, Dsc1a, Dsc1b
          Chromosome Location:
          Chr.18: 20083471 - 20114773 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00496525_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Dsc1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001291804.1 NP_001278733.1 6 - 7 988 100
          NM_013504.5 NP_038532.3 6 - 7 988 100
          GenBank mRNA AK036893.1 - 6 - 7 874 100
          AK079441.1 - 6 - 7 990 100
          AK081826.1 - 6 - 7 896 100
          BC109161.1 - 6 - 7 825 40047883 100
          BC109162.1 - 6 - 7 825 100
          X97986.1 - 6 - 7 922 100

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1800001045.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Dsc1

          Panther Classification:

          Molecular Function -

          cadherin

          Gene Ontology Categories:

          Function(s) Process(es)

          cell adhesion
          homophilic cell adhesion via plasma membrane adhesion molecules
          calcium ion binding
          protein binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2157735 AK079441.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2157735 NM_013504.4 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 904036 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 763873 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 504752 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 51991 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 457202 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 620748 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 377286 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 138126 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 412612 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 517172 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 367177 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 205454 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 334356 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 790364 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 659838 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 777217 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 761670 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 175562 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 81924 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P108575 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P154709 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2157735 AK079441.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2157735 NM_013504.4 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P437166 X97986.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P437166 AK081826.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P437166 NM_013504.4 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P437166 AK079441.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P03424329 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P03424347 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P108575 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P154709 - -
          Whole Mouse Genome G4122A A_51_P437166 X97986.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P437166 AK081826.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P437166 NM_013504.4 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P437166 AK079441.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P123179 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P154709 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P108575 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P03424347 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14059436 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13800516 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13800524 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14059427 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14059438 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13800527 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31537639 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31537689 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31537745 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31537596 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31537662 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31537692 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31537647 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31537778 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31537602 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31537748 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31537629 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31537589 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31537650 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31537706 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31537682 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31537624 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31537718 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31537712 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31537792 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31537592 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31537599 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31537660 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31537709 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31537606 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31537643 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31537784 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31537732 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31537736 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31537597 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31537773 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31537729 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31537775 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31537591 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31537655 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31537668 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31537674 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31537699 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31537725 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31537654 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31537767 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31537691 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31537672 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31537644 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31537733 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31537789 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31537687 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31537626 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31537798 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31537593 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31537790 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31537721 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31537787 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31537696 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31537636 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31537610 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31537613 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31537594 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31537642 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31537637 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31537697 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31537701 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31537795 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31537710 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31537759 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31537783 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31537785 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31537764 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31537693 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31537780 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31537676 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31537630 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31537604 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31537679 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31537652 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31537617 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31537727 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31537722 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31537670 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31537605 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31537666 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31537743 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31537756 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31537665 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31537608 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31537678 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31537738 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31537702 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31537771 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31537758 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31537754 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31537658 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31537685 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31537731 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31537690 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31537704 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31537724 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31537614 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31537700 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31537760 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31537750 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31537695 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31537746 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31537621 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31537633 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31537799 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10457715 - -
          Affymetrix Mouse Gene 1.0 ST Array 10457728 - -
          Affymetrix Mouse Gene 1.0 ST Array 10457721 - -
          Affymetrix Mouse Gene 1.0 ST Array 10457722 - -
          Affymetrix Mouse Gene 1.0 ST Array 10457719 - -
          Affymetrix Mouse Gene 1.0 ST Array 10457726 - -
          Affymetrix Mouse Gene 1.0 ST Array 10457713 - -
          Affymetrix Mouse Gene 1.0 ST Array 10457712 - -
          Affymetrix Mouse Gene 1.0 ST Array 10457717 - -
          Affymetrix Mouse Gene 1.0 ST Array 10457716 - -
          Affymetrix Mouse Gene 1.0 ST Array 10457710 - -
          Affymetrix Mouse Gene 1.0 ST Array 10457725 - -
          Affymetrix Mouse Gene 1.0 ST Array 10457708 - -
          Affymetrix Mouse Gene 1.0 ST Array 10457714 - -
          Affymetrix Mouse Gene 1.0 ST Array 10457711 - -
          Affymetrix Mouse Gene 1.0 ST Array 10457724 - -
          Affymetrix Mouse Gene 1.0 ST Array 10457727 - -
          Affymetrix Mouse Gene 1.0 ST Array 10457720 - -
          Affymetrix Mouse Gene 1.0 ST Array 10457709 - -
          Affymetrix Mouse Gene 1.0 ST Array 10457718 - -
          Affymetrix Mouse Gene 1.0 ST Array 10457723 - -
          Affymetrix HT MG-430 PM Array Plate 1421460_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10457707 - -
          Mouse Genome 430 2.0 Array 1421460_PM_at - -
          Mouse Genome 430 2.0 Array 1421460_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 13505_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 13505_at - -
          Affymetrix Mouse Clariom S Array PSR1800007723.mm.1 NM_013504.5 Refseq
          Affymetrix Mouse Clariom S Array PSR1800007723.mm.1 TC1800001045.mm.1 MTA TC
          Mouse Expression Set 430 1421460_PM_at - -
          Mouse Expression Set 430 1421460_at - -
          Murine Genome U74v2 Set 100760_PM_at - -
          Murine Genome U74v2 Set 100760_at - -
          Mouse Genome 430A 2.0 Array 1421460_PM_at - -
          Mouse Genome 430A 2.0 Array 1421460_at - -
          Affymetrix HT Mouse Genome 430A Array 1421460_at - -
          Affymetrix HT Mouse Genome 430A Array 1421460_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 434440 - -
          ABI Mouse Genome Survey Microarray 441020 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 6130400 - -
          MouseWG-6 v2 Expression BeadChip 106510086 - -
          Illumina mouse-6 v1.0 expression beadchip 6130400 - -
          Illumina mouse-6 v1.0 expression beadchip 106510086 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006130400 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2723533 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2723533 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1234954 - -
          mouse-6 v1.1 (Illumina) scl013505.1_42-S - -
          Illumina Sentrix mouse-6.v1 bead array 106510086 - -
          Illumina Sentrix mouse-6.v1 bead array 6130400 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl013505.1_42-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6130400 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 106510086 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2723533 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1234954 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2723533 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline