Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00496595_m1
          See other GRCC10 GE Assays ›
          Gene Symbol:
          Grcc10
          Gene Name:
          gene rich cluster, C10 gene
          Gene Aliases:
          2310033H05Rik, C10
          Chromosome Location:
          Chr.6: 124739183 - 124741079 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00496595_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Grcc10
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_013535.1 NP_038563.1 2 - 3 343 105
          GenBank mRNA AK009591.1 - 2 - 3 270 105
          BC005683.1 - 2 - 3 322 3154555 105

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0600003603.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Grcc10

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24816734 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24816729 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24816733 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24816731 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24816728 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24816730 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1978618 AK009591.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1978618 AK013899.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1978618 BC005683.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1978618 NM_013535.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 851213 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P178353 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1978618 AK009591.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1978618 AK013899.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1978618 BC005683.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1978618 NM_013535.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24816733 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24816734 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24816742 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24816736 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24816722 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24816738 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24816731 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24816729 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24816725 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24816726 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24816740 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24816737 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24816739 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24816723 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P178353 - -
          Whole Mouse Genome G4122A A_51_P320114 BC005683.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P320114 NM_013535.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P130893 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P178353 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05256438 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05256435 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05831627 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05256439 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05686757 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05831630 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05686756 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24816733 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24816730 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24816729 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24816734 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24816728 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24816731 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24816670 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24816737 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24816658 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24816796 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24816727 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24816683 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24816739 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24816697 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24816663 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24816677 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24816667 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24816726 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24816718 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24816686 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24816666 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24816700 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24816657 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24816736 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24816720 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24816693 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24816734 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24816800 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24816733 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24816681 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24816673 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24816695 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24816798 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24816675 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24816725 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24816740 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24816674 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24816679 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24816731 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24816660 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24816656 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24816671 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24816729 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24816654 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24816733 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05686757 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05109516 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24816724 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05405482 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24816725 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24816729 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24816726 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24816736 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05256438 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24816740 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05831628 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24816742 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05387415 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24816731 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10547792 - -
          Affymetrix Mouse Gene 1.0 ST Array 10547790 - -
          Affymetrix Mouse Gene 1.0 ST Array 10547791 - -
          Affymetrix HT MG-430 PM Array Plate 1416522_PM_a_at - -
          Affymetrix HT MG-430 PM Array Plate 1429782_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10547789 - -
          Mouse Genome 430 2.0 Array 1429782_at - -
          Mouse Genome 430 2.0 Array 1416522_a_at - -
          Mouse Genome 430 2.0 Array 1416522_PM_a_at - -
          Mouse Genome 430 2.0 Array 1429782_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 14790_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 14790_at - -
          Affymetrix Mouse Clariom S Array PSR0600027674.mm.1 TC0600003603.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600027674.mm.1 NM_013535.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600027674.mm.1 NM_013535.1 Refseq
          Mouse Expression Set 430 1429782_PM_at - -
          Mouse Expression Set 430 1416522_a_at - -
          Mouse Expression Set 430 1416522_PM_a_at - -
          Mouse Expression Set 430 1429782_at - -
          Murine Genome U74v2 Set 94206_PM_at - -
          Murine Genome U74v2 Set 94206_at - -
          Mouse Genome 430A 2.0 Array 1429782_at - -
          Mouse Genome 430A 2.0 Array 1416522_PM_a_at - -
          Mouse Genome 430A 2.0 Array 1429782_PM_at - -
          Mouse Genome 430A 2.0 Array 1416522_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1429782_at - -
          Affymetrix HT Mouse Genome 430A Array 1416522_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1416522_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1429782_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 457775 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 3840279 - -
          Illumina mouse-6 v1.0 expression beadchip 3840279 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003840279 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2722678 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2722678 - -
          mouse-6 v1.1 (Illumina) scl014790.2_94-S - -
          Illumina Sentrix mouse-6.v1 bead array 3840279 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl014790.2_94-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3840279 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2722678 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2722678 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl014790.2_94-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline