| Array Name |
Probe IDs |
Targets |
Target Type |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2036472 |
NM_001161832.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2036472 |
AF072546.1 |
mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
355013 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
292638 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
297045 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
376142 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
647785 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
84383 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
532585 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
700165 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
881580 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
63729 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
502809 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
649099 |
AK032014.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
649099 |
AK035460.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
797896 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
96155 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
203436 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
401386 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
771191 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
338803 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
973843 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
838012 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
871719 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
548893 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
667268 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
322977 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
706852 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
504635 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
612802 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
900319 |
BC065803.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
900319 |
NM_178186.2 |
refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
780869 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
811073 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
950233 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
107465 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
112235 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
138498 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
167243 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
105916 |
- |
- |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2036472 |
NM_001161832.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2036472 |
AF072546.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P261412 |
BC119261.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P261412 |
BC119287.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P261412 |
NM_001161832.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P261412 |
AF072546.1 |
mouse_transcriptome |